Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124810083-124810356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARB1
Entrez Gene ID
949
Gene Name
scavenger receptor class B member 1
Gene Aliases
CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1
UniGene
Hs.731377
Gene Chromosome Location
Chr.12: 124777628-124863973 on Build GRCh38
Transcripts
NM_001082959.1 NM_005505.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398502 c.832G>T p.E278* Chr.12: 124810184-124810184 on Build GRCh38
4860856 c.708G>A p.E236E Chr.12: 124810197-124810197 on Build GRCh38
6037640 c.702G>A p.S234S Chr.12: 124810203-124810203 on Build GRCh38
6037639 c.813G>A p.S271S Chr.12: 124810203-124810203 on Build GRCh38
692396 c.819G>A p.E273E Chr.12: 124810197-124810197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142176147 Chr.12: 124810205-124810205 on Build GRCh38
rs771957199 Chr.12: 124810175-124810175 on Build GRCh38
rs753314676 Chr.12: 124810220-124810220 on Build GRCh38
rs777036596 Chr.12: 124810272-124810272 on Build GRCh38
rs779498997 Chr.12: 124810217-124810217 on Build GRCh38
rs200952914 Chr.12: 124810154-124810154 on Build GRCh38
rs966110622 Chr.12: 124810330-124810330 on Build GRCh38
rs774310843 Chr.12: 124810166-124810166 on Build GRCh38
rs751116119 Chr.12: 124810313-124810314 on Build GRCh38
rs763215766 Chr.12: 124810204-124810204 on Build GRCh38
rs572531355 Chr.12: 124810167-124810167 on Build GRCh38
rs768611337 Chr.12: 124810162-124810162 on Build GRCh38
rs748180440 Chr.12: 124810233-124810233 on Build GRCh38
rs775858958 Chr.12: 124810292-124810292 on Build GRCh38
rs771694795 Chr.12: 124810242-124810242 on Build GRCh38
rs757862285 Chr.12: 124810152-124810152 on Build GRCh38
rs374077998 Chr.12: 124810294-124810294 on Build GRCh38
rs759334503 Chr.12: 124810203-124810203 on Build GRCh38
rs201357313 Chr.12: 124810271-124810271 on Build GRCh38
rs763522774 Chr.12: 124810293-124810293 on Build GRCh38
rs565034153 Chr.12: 124810227-124810227 on Build GRCh38
rs757068361 Chr.12: 124810158-124810158 on Build GRCh38
rs542972166 Chr.12: 124810122-124810122 on Build GRCh38
rs776877098 Chr.12: 124810192-124810192 on Build GRCh38
rs760953431 Chr.12: 124810182-124810182 on Build GRCh38
rs539778772 Chr.12: 124810108-124810108 on Build GRCh38
rs749446809 Chr.12: 124810242-124810242 on Build GRCh38
rs765896844 Chr.12: 124810227-124810228 on Build GRCh38
rs762203805 Chr.12: 124810322-124810322 on Build GRCh38
rs188756468 Chr.12: 124810298-124810298 on Build GRCh38
rs773298594 Chr.12: 124810181-124810181 on Build GRCh38
rs765811290 Chr.12: 124810201-124810201 on Build GRCh38
rs536512405 Chr.12: 124810152-124810152 on Build GRCh38
rs759669126 Chr.12: 124810287-124810287 on Build GRCh38
rs924703004 Chr.12: 124810170-124810170 on Build GRCh38
rs765992920 Chr.12: 124810333-124810333 on Build GRCh38
rs755777295 Chr.12: 124810216-124810216 on Build GRCh38
rs369487060 Chr.12: 124810222-124810222 on Build GRCh38
rs201068540 Chr.12: 124810291-124810291 on Build GRCh38
rs957503689 Chr.12: 124810180-124810180 on Build GRCh38
rs749977931 Chr.12: 124810213-124810213 on Build GRCh38
rs547400313 Chr.12: 124810185-124810185 on Build GRCh38
rs868130307 Chr.12: 124810244-124810244 on Build GRCh38
rs376944735 Chr.12: 124810157-124810157 on Build GRCh38
rs377441377 Chr.12: 124810228-124810228 on Build GRCh38
rs369722279 Chr.12: 124810200-124810200 on Build GRCh38
rs777634591 Chr.12: 124810252-124810252 on Build GRCh38

View Full Product Details