Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31927912-31928179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NM_001282459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1443404 c.88A>G p.I30V Chr.6: 31927996-31927996 on Build GRCh38
451225 c.256C>T p.P86S Chr.6: 31928164-31928164 on Build GRCh38
3921404 c.174C>T p.S58S Chr.6: 31928082-31928082 on Build GRCh38
3921403 c.135C>G p.S45R Chr.6: 31928043-31928043 on Build GRCh38
3624851 c.256+9C>T p.? Chr.6: 31928173-31928173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779595202 Chr.6: 31928138-31928138 on Build GRCh38
rs750215781 Chr.6: 31928111-31928111 on Build GRCh38
rs377750767 Chr.6: 31928012-31928012 on Build GRCh38
rs894441537 Chr.6: 31927943-31927943 on Build GRCh38
rs961046350 Chr.6: 31928084-31928084 on Build GRCh38
rs769462258 Chr.6: 31927994-31927994 on Build GRCh38
rs769289608 Chr.6: 31928009-31928009 on Build GRCh38
rs751453267 Chr.6: 31928097-31928097 on Build GRCh38
rs756248699 Chr.6: 31927967-31927967 on Build GRCh38
rs745474379 Chr.6: 31927990-31927990 on Build GRCh38
rs751363286 Chr.6: 31927986-31927986 on Build GRCh38
rs200095096 Chr.6: 31927981-31927981 on Build GRCh38
rs547881286 Chr.6: 31928153-31928153 on Build GRCh38
rs760856379 Chr.6: 31928076-31928076 on Build GRCh38
rs771656533 Chr.6: 31928127-31928128 on Build GRCh38
rs754668308 Chr.6: 31928154-31928154 on Build GRCh38
rs774919002 Chr.6: 31927935-31927935 on Build GRCh38
rs572850142 Chr.6: 31927976-31927976 on Build GRCh38
rs376686530 Chr.6: 31928115-31928115 on Build GRCh38
rs780894869 Chr.6: 31928110-31928110 on Build GRCh38
rs201336507 Chr.6: 31927989-31927989 on Build GRCh38
rs561819885 Chr.6: 31927998-31927998 on Build GRCh38
rs146464977 Chr.6: 31928092-31928092 on Build GRCh38
rs779783858 Chr.6: 31928125-31928125 on Build GRCh38
rs137902889 Chr.6: 31928126-31928126 on Build GRCh38
rs772970793 Chr.6: 31928140-31928141 on Build GRCh38
rs753987052 Chr.6: 31928082-31928082 on Build GRCh38
rs750561586 Chr.6: 31927951-31927951 on Build GRCh38
rs757110747 Chr.6: 31928109-31928109 on Build GRCh38
rs1008127965 Chr.6: 31928073-31928073 on Build GRCh38
rs766410942 Chr.6: 31927968-31927968 on Build GRCh38
rs764034564 Chr.6: 31928093-31928093 on Build GRCh38
rs762200436 Chr.6: 31927946-31927946 on Build GRCh38
rs200412106 Chr.6: 31928000-31928000 on Build GRCh38
rs201909990 Chr.6: 31928029-31928029 on Build GRCh38
rs773595474 Chr.6: 31928066-31928066 on Build GRCh38
rs997913124 Chr.6: 31928075-31928075 on Build GRCh38
rs566638128 Chr.6: 31928136-31928136 on Build GRCh38
rs748642938 Chr.6: 31928145-31928145 on Build GRCh38
rs1057441905 Chr.6: 31927938-31927938 on Build GRCh38
rs768231128 Chr.6: 31928137-31928137 on Build GRCh38
rs772283614 Chr.6: 31928063-31928063 on Build GRCh38
rs766612461 Chr.6: 31928081-31928081 on Build GRCh38

View Full Product Details