Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66081656-66081909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5716568 c.63G>A p.M21I Chr.7: 66081853-66081853 on Build GRCh38
4838079 c.90C>T p.Y30Y Chr.7: 66081880-66081880 on Build GRCh38
1330553 c.93C>T p.D31D Chr.7: 66081883-66081883 on Build GRCh38
3882062 c.76G>A p.A26T Chr.7: 66081866-66081866 on Build GRCh38
7203941 c.108G>A p.E36E Chr.7: 66081898-66081898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760059034 Chr.7: 66081862-66081862 on Build GRCh38
rs754995756 Chr.7: 66081881-66081881 on Build GRCh38
rs985659014 Chr.7: 66081715-66081715 on Build GRCh38
rs201082875 Chr.7: 66081799-66081799 on Build GRCh38
rs549560616 Chr.7: 66081790-66081790 on Build GRCh38
rs752589473 Chr.7: 66081704-66081704 on Build GRCh38
rs375596811 Chr.7: 66081745-66081745 on Build GRCh38
rs761046404 Chr.7: 66081867-66081867 on Build GRCh38
rs376108729 Chr.7: 66081752-66081752 on Build GRCh38
rs917367197 Chr.7: 66081725-66081725 on Build GRCh38
rs774392615 Chr.7: 66081852-66081852 on Build GRCh38
rs775839623 Chr.7: 66081866-66081866 on Build GRCh38
rs767920084 Chr.7: 66081766-66081766 on Build GRCh38
rs753013599 Chr.7: 66081768-66081768 on Build GRCh38
rs760005936 Chr.7: 66081765-66081765 on Build GRCh38
rs571705199 Chr.7: 66081827-66081827 on Build GRCh38
rs529417509 Chr.7: 66081708-66081708 on Build GRCh38
rs145138923 Chr.7: 66081825-66081825 on Build GRCh38
rs749918615 Chr.7: 66081792-66081792 on Build GRCh38
rs142077823 Chr.7: 66081889-66081889 on Build GRCh38
rs910643561 Chr.7: 66081850-66081850 on Build GRCh38
rs1022405993 Chr.7: 66081744-66081744 on Build GRCh38
rs149108910 Chr.7: 66081885-66081885 on Build GRCh38
rs968294351 Chr.7: 66081757-66081757 on Build GRCh38
rs751062108 Chr.7: 66081880-66081880 on Build GRCh38
rs763073054 Chr.7: 66081839-66081839 on Build GRCh38
rs901715532 Chr.7: 66081884-66081884 on Build GRCh38
rs757988466 Chr.7: 66081796-66081796 on Build GRCh38
rs771082435 Chr.7: 66081848-66081848 on Build GRCh38
rs781154096 Chr.7: 66081818-66081818 on Build GRCh38
rs757952889 Chr.7: 66081879-66081879 on Build GRCh38
rs115069006 Chr.7: 66081868-66081868 on Build GRCh38
rs147614617 Chr.7: 66081695-66081695 on Build GRCh38
rs1048954249 Chr.7: 66081864-66081864 on Build GRCh38
rs773425505 Chr.7: 66081831-66081831 on Build GRCh38
rs768690501 Chr.7: 66081813-66081813 on Build GRCh38
rs930628311 Chr.7: 66081838-66081838 on Build GRCh38
rs370260834 Chr.7: 66081865-66081865 on Build GRCh38
rs572094840 Chr.7: 66081842-66081842 on Build GRCh38
rs564735357 Chr.7: 66081824-66081824 on Build GRCh38
rs200194982 Chr.7: 66081810-66081810 on Build GRCh38
rs763823679 Chr.7: 66081856-66081856 on Build GRCh38

View Full Product Details