Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48600759-48601028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UQCRC1
Entrez Gene ID
7384
Gene Name
ubiquinol-cytochrome c reductase core protein I
Gene Aliases
D3S3191, QCR1, UQCR1
UniGene
Hs.119251
Gene Chromosome Location
Chr.3: 48598999-48609665 on Build GRCh38
Transcripts
NM_003365.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1231859 c.1028C>T p.T343I Chr.3: 48600779-48600779 on Build GRCh38
3595028 c.936C>T p.G312G Chr.3: 48601005-48601005 on Build GRCh38
7301184 c.933C>T p.I311I Chr.3: 48601008-48601008 on Build GRCh38
3373214 c.949A>G p.T317A Chr.3: 48600992-48600992 on Build GRCh38
1581911 c.923A>G p.N308S Chr.3: 48601018-48601018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776670206 Chr.3: 48600886-48600886 on Build GRCh38
rs551263187 Chr.3: 48600838-48600838 on Build GRCh38
rs746144966 Chr.3: 48600876-48600876 on Build GRCh38
rs903826336 Chr.3: 48600874-48600874 on Build GRCh38
rs746851503 Chr.3: 48600882-48600882 on Build GRCh38
rs369530651 Chr.3: 48600887-48600887 on Build GRCh38
rs963843477 Chr.3: 48600893-48600893 on Build GRCh38
rs772865589 Chr.3: 48600946-48600946 on Build GRCh38
rs753836240 Chr.3: 48600843-48600843 on Build GRCh38
rs755099413 Chr.3: 48600986-48600988 on Build GRCh38
rs780276583 Chr.3: 48600988-48600988 on Build GRCh38
rs536141675 Chr.3: 48600787-48600787 on Build GRCh38
rs768457004 Chr.3: 48600885-48600885 on Build GRCh38
rs539018849 Chr.3: 48600957-48600957 on Build GRCh38
rs373227178 Chr.3: 48600978-48600978 on Build GRCh38
rs1056556879 Chr.3: 48600811-48600811 on Build GRCh38
rs759969765 Chr.3: 48600792-48600792 on Build GRCh38
rs759945732 Chr.3: 48600966-48600966 on Build GRCh38
rs758630275 Chr.3: 48600877-48600877 on Build GRCh38
rs950762023 Chr.3: 48600807-48600807 on Build GRCh38
rs557741559 Chr.3: 48600959-48600959 on Build GRCh38
rs375097385 Chr.3: 48600793-48600793 on Build GRCh38
rs184725145 Chr.3: 48600961-48600961 on Build GRCh38
rs756541768 Chr.3: 48600813-48600813 on Build GRCh38
rs939548148 Chr.3: 48600806-48600806 on Build GRCh38
rs780978963 Chr.3: 48601000-48601000 on Build GRCh38
rs779100134 Chr.3: 48600856-48600856 on Build GRCh38
rs955256071 Chr.3: 48600903-48600903 on Build GRCh38
rs750241389 Chr.3: 48600930-48600931 on Build GRCh38
rs779757018 Chr.3: 48600880-48600880 on Build GRCh38
rs761865695 Chr.3: 48600963-48600963 on Build GRCh38
rs569558027 Chr.3: 48600932-48600932 on Build GRCh38
rs754870725 Chr.3: 48600992-48600992 on Build GRCh38
rs1008102480 Chr.3: 48600910-48600910 on Build GRCh38
rs555118861 Chr.3: 48600977-48600977 on Build GRCh38
rs762631105 Chr.3: 48600947-48600947 on Build GRCh38
rs751282347 Chr.3: 48600991-48600991 on Build GRCh38
rs757395133 Chr.3: 48600851-48600851 on Build GRCh38
rs1044117778 Chr.3: 48600868-48600868 on Build GRCh38
rs770532906 Chr.3: 48600875-48600875 on Build GRCh38
rs753761633 Chr.3: 48600870-48600870 on Build GRCh38
rs142936717 Chr.3: 48600960-48600960 on Build GRCh38
rs757234390 Chr.3: 48600875-48600876 on Build GRCh38
rs764986255 Chr.3: 48600874-48600875 on Build GRCh38
rs1012297268 Chr.3: 48600815-48600815 on Build GRCh38
rs145869559 Chr.3: 48601007-48601007 on Build GRCh38
rs753120561 Chr.3: 48600800-48600800 on Build GRCh38
rs201707020 Chr.3: 48600987-48600987 on Build GRCh38
rs769861368 Chr.3: 48601008-48601008 on Build GRCh38
rs377152335 Chr.3: 48600962-48600962 on Build GRCh38

View Full Product Details