Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181579019-181579292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
UniGene
Hs.437444
Gene Chromosome Location
Chr.1: 181317712-181808084 on Build GRCh38
Transcripts
NM_000721.3 NM_001205293.1 NM_001205294.1 XM_017002243.1 XM_017002244.1 XM_017002245.1 XM_017002246.1 XM_017002247.1 XM_017002248.1 XM_017002249.1 XM_017002250.1 XM_017002251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7075085 c.525T>A p.I175I Chr.1: 181579127-181579127 on Build GRCh38
5758058 c.593T>A p.L198* Chr.1: 181579195-181579195 on Build GRCh38
6258386 c.605G>T p.C202F Chr.1: 181579207-181579207 on Build GRCh38
6059827 c.507G>T p.M169I Chr.1: 181579109-181579109 on Build GRCh38
5758057 c.740T>A p.L247* Chr.1: 181579195-181579195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760987341 Chr.1: 181579091-181579091 on Build GRCh38
rs768321184 Chr.1: 181579250-181579250 on Build GRCh38
rs751937009 Chr.1: 181579263-181579263 on Build GRCh38
rs113730525 Chr.1: 181579056-181579056 on Build GRCh38
rs770708783 Chr.1: 181579259-181579259 on Build GRCh38
rs755117584 Chr.1: 181579190-181579190 on Build GRCh38
rs778934511 Chr.1: 181579198-181579198 on Build GRCh38
rs747273664 Chr.1: 181579233-181579233 on Build GRCh38
rs774493647 Chr.1: 181579044-181579044 on Build GRCh38
rs758787022 Chr.1: 181579215-181579215 on Build GRCh38
rs966372276 Chr.1: 181579045-181579045 on Build GRCh38
rs17443123 Chr.1: 181579205-181579205 on Build GRCh38
rs371977742 Chr.1: 181579085-181579085 on Build GRCh38
rs375614479 Chr.1: 181579094-181579094 on Build GRCh38
rs766455994 Chr.1: 181579092-181579092 on Build GRCh38
rs771862571 Chr.1: 181579268-181579268 on Build GRCh38
rs769497204 Chr.1: 181579243-181579243 on Build GRCh38
rs546161068 Chr.1: 181579231-181579231 on Build GRCh38
rs985328341 Chr.1: 181579082-181579082 on Build GRCh38
rs955373152 Chr.1: 181579070-181579070 on Build GRCh38
rs564826658 Chr.1: 181579266-181579266 on Build GRCh38
rs748940167 Chr.1: 181579246-181579246 on Build GRCh38

View Full Product Details