Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 88990997-88991270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf47
Entrez Gene ID
286223
Gene Name
chromosome 9 open reading frame 47
Gene Aliases
C9orf108, bA791O21.3
UniGene
Hs.585118
Gene Chromosome Location
Chr.9: 88990863-88996142 on Build GRCh38
Transcripts
NM_001001938.3 NM_001142413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3927186 c.121C>T p.P41S Chr.9: 88991116-88991116 on Build GRCh38
4188618 c.72_76delGCTGT p.L25fs*35 Chr.9: 88991067-88991071 on Build GRCh38
4626493 c.208G>T p.E70* Chr.9: 88991203-88991203 on Build GRCh38
4626494 c.239G>T p.R80M Chr.9: 88991234-88991234 on Build GRCh38
3659389 c.8G>A p.R3K Chr.9: 88991003-88991003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004131740 Chr.9: 88991252-88991252 on Build GRCh38
rs774721827 Chr.9: 88991196-88991196 on Build GRCh38
rs755324673 Chr.9: 88991127-88991127 on Build GRCh38
rs779987121 Chr.9: 88991230-88991230 on Build GRCh38
rs768319593 Chr.9: 88991078-88991078 on Build GRCh38
rs887117157 Chr.9: 88991226-88991226 on Build GRCh38
rs140930200 Chr.9: 88991022-88991022 on Build GRCh38
rs778761127 Chr.9: 88991062-88991062 on Build GRCh38
rs762573987 Chr.9: 88991191-88991191 on Build GRCh38
rs542361091 Chr.9: 88991214-88991214 on Build GRCh38
rs537215480 Chr.9: 88991129-88991129 on Build GRCh38
rs552447519 Chr.9: 88991163-88991163 on Build GRCh38
rs767757524 Chr.9: 88991201-88991201 on Build GRCh38
rs772824749 Chr.9: 88991173-88991173 on Build GRCh38
rs769608875 Chr.9: 88991169-88991169 on Build GRCh38
rs759883642 Chr.9: 88991197-88991197 on Build GRCh38
rs148798807 Chr.9: 88991070-88991070 on Build GRCh38
rs946942025 Chr.9: 88991148-88991148 on Build GRCh38
rs943672219 Chr.9: 88991218-88991218 on Build GRCh38
rs532405326 Chr.9: 88991125-88991125 on Build GRCh38
rs369697077 Chr.9: 88991124-88991124 on Build GRCh38
rs772921523 Chr.9: 88991110-88991110 on Build GRCh38
rs1024014585 Chr.9: 88991115-88991115 on Build GRCh38
rs767041180 Chr.9: 88991116-88991116 on Build GRCh38
rs756414401 Chr.9: 88991132-88991132 on Build GRCh38
rs145434754 Chr.9: 88991066-88991066 on Build GRCh38
rs905469386 Chr.9: 88991171-88991171 on Build GRCh38
rs150059331 Chr.9: 88991048-88991048 on Build GRCh38
rs760866323 Chr.9: 88991213-88991213 on Build GRCh38
rs748214982 Chr.9: 88991058-88991058 on Build GRCh38
rs781292375 Chr.9: 88991049-88991049 on Build GRCh38
rs763522209 Chr.9: 88991113-88991113 on Build GRCh38
rs749421961 Chr.9: 88991142-88991142 on Build GRCh38
rs924291016 Chr.9: 88991101-88991101 on Build GRCh38
rs756149226 Chr.9: 88991061-88991061 on Build GRCh38
rs557568274 Chr.9: 88991143-88991143 on Build GRCh38
rs764828613 Chr.9: 88991105-88991105 on Build GRCh38
rs760435447 Chr.9: 88991033-88991035 on Build GRCh38
rs34528137 Chr.9: 88991219-88991219 on Build GRCh38
rs139376565 Chr.9: 88991030-88991030 on Build GRCh38
rs777801468 Chr.9: 88991138-88991138 on Build GRCh38
rs971035529 Chr.9: 88991133-88991133 on Build GRCh38
rs781100018 Chr.9: 88991157-88991157 on Build GRCh38
rs753170629 Chr.9: 88991207-88991207 on Build GRCh38
rs746751572 Chr.9: 88991145-88991145 on Build GRCh38
rs776153271 Chr.9: 88991084-88991084 on Build GRCh38
rs761330671 Chr.9: 88991100-88991100 on Build GRCh38
rs768350723 Chr.9: 88991149-88991149 on Build GRCh38
rs990295471 Chr.9: 88991184-88991184 on Build GRCh38
rs780002221 Chr.9: 88991144-88991144 on Build GRCh38
rs867630503 Chr.9: 88991221-88991221 on Build GRCh38
rs35286411 Chr.9: 88991227-88991228 on Build GRCh38
rs775246429 Chr.9: 88991069-88991069 on Build GRCh38
rs751441912 Chr.9: 88991232-88991232 on Build GRCh38
rs201570852 Chr.9: 88991195-88991195 on Build GRCh38
rs376789769 Chr.9: 88991220-88991220 on Build GRCh38

View Full Product Details