Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29744097-29744326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf54
Entrez Gene ID
283897
Gene Name
chromosome 16 open reading frame 54
Gene Aliases
-
UniGene
Hs.331095
Gene Chromosome Location
Chr.16: 29742463-29746019 on Build GRCh38
Transcripts
NM_175900.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
396937 c.627G>T p.Q209H Chr.16: 29744325-29744325 on Build GRCh38
213684 c.632C>G p.S211* Chr.16: 29744320-29744320 on Build GRCh38
3508568 c.642G>A p.W214* Chr.16: 29744310-29744310 on Build GRCh38
4666974 c.646C>T p.R216C Chr.16: 29744306-29744306 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762218040 Chr.16: 29744289-29744289 on Build GRCh38
rs775549533 Chr.16: 29744305-29744305 on Build GRCh38
rs749333124 Chr.16: 29744288-29744288 on Build GRCh38
rs758114934 Chr.16: 29744253-29744253 on Build GRCh38
rs899766599 Chr.16: 29744225-29744225 on Build GRCh38
rs563935958 Chr.16: 29744217-29744217 on Build GRCh38
rs1015561894 Chr.16: 29744224-29744224 on Build GRCh38
rs545636576 Chr.16: 29744208-29744208 on Build GRCh38
rs958142216 Chr.16: 29744286-29744286 on Build GRCh38
rs778501279 Chr.16: 29744243-29744243 on Build GRCh38
rs756785016 Chr.16: 29744242-29744242 on Build GRCh38
rs748977284 Chr.16: 29744175-29744175 on Build GRCh38
rs931490524 Chr.16: 29744138-29744138 on Build GRCh38
rs750050863 Chr.16: 29744250-29744250 on Build GRCh38
rs770135296 Chr.16: 29744268-29744268 on Build GRCh38
rs1007864508 Chr.16: 29744205-29744205 on Build GRCh38
rs778439639 Chr.16: 29744280-29744280 on Build GRCh38
rs141257864 Chr.16: 29744176-29744176 on Build GRCh38
rs771166622 Chr.16: 29744296-29744296 on Build GRCh38
rs774614027 Chr.16: 29744297-29744297 on Build GRCh38
rs746359702 Chr.16: 29744251-29744251 on Build GRCh38
rs898697809 Chr.16: 29744137-29744137 on Build GRCh38
rs201665846 Chr.16: 29744299-29744299 on Build GRCh38
rs1057422787 Chr.16: 29744133-29744133 on Build GRCh38
rs112310246 Chr.16: 29744255-29744255 on Build GRCh38
rs774723249 Chr.16: 29744215-29744215 on Build GRCh38
rs1047280460 Chr.16: 29744161-29744161 on Build GRCh38
rs748615663 Chr.16: 29744260-29744260 on Build GRCh38
rs753289692 Chr.16: 29744236-29744236 on Build GRCh38

View Full Product Details