Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40389837-40390064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNSTRN
Entrez Gene ID
90417
Gene Name
kinetochore localized astrin/SPAG5 binding protein
Gene Aliases
C15orf23, HSD11, SKAP, TRAF4AF1
UniGene
Hs.525796
Gene Chromosome Location
Chr.15: 40382721-40394288 on Build GRCh38
Transcripts
NM_001142761.1 NM_001142762.1 NM_033286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6697609 c.641G>A p.R214Q Chr.15: 40389885-40389885 on Build GRCh38
6293628 c.655G>T p.A219S Chr.15: 40389899-40389899 on Build GRCh38
7374298 c.622G>C p.E208Q Chr.15: 40389866-40389866 on Build GRCh38
7374297 c.622G>C p.E208Q Chr.15: 40389866-40389866 on Build GRCh38
6293629 c.655G>T p.A219S Chr.15: 40389899-40389899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746335299 Chr.15: 40389980-40389980 on Build GRCh38
rs771409374 Chr.15: 40389866-40389866 on Build GRCh38
rs749906792 Chr.15: 40389888-40389888 on Build GRCh38
rs761530012 Chr.15: 40389884-40389884 on Build GRCh38
rs371547856 Chr.15: 40389876-40389876 on Build GRCh38
rs754860618 Chr.15: 40389922-40389922 on Build GRCh38
rs994884480 Chr.15: 40389859-40389859 on Build GRCh38
rs201422345 Chr.15: 40389885-40389885 on Build GRCh38
rs191518976 Chr.15: 40389897-40389897 on Build GRCh38
rs749321382 Chr.15: 40389968-40389968 on Build GRCh38
rs770908025 Chr.15: 40389970-40389970 on Build GRCh38
rs991141684 Chr.15: 40389907-40389907 on Build GRCh38
rs780944766 Chr.15: 40389936-40389936 on Build GRCh38
rs924552599 Chr.15: 40390024-40390024 on Build GRCh38
rs934462857 Chr.15: 40390034-40390034 on Build GRCh38
rs1042959658 Chr.15: 40390021-40390021 on Build GRCh38
rs61739399 Chr.15: 40389878-40389878 on Build GRCh38
rs751449930 Chr.15: 40389918-40389918 on Build GRCh38
rs182422591 Chr.15: 40389967-40389967 on Build GRCh38
rs963577856 Chr.15: 40389863-40389863 on Build GRCh38
rs151241303 Chr.15: 40389919-40389919 on Build GRCh38
rs959832551 Chr.15: 40389899-40389899 on Build GRCh38
rs779438003 Chr.15: 40389873-40389873 on Build GRCh38
rs774580885 Chr.15: 40389869-40389869 on Build GRCh38
rs756280416 Chr.15: 40389953-40389953 on Build GRCh38
rs766275025 Chr.15: 40389898-40389898 on Build GRCh38
rs947276904 Chr.15: 40389988-40389988 on Build GRCh38
rs776468154 Chr.15: 40389880-40389880 on Build GRCh38
rs774978842 Chr.15: 40389973-40389973 on Build GRCh38
rs748399580 Chr.15: 40389943-40389943 on Build GRCh38

View Full Product Details