Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127706464-127706705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR33
Entrez Gene ID
55339
Gene Name
WD repeat domain 33
Gene Aliases
NET14, WDC146
UniGene
Hs.554831
Gene Chromosome Location
Chr.2: 127704234-127811187 on Build GRCh38
Transcripts
NM_018383.4 XM_005263697.2 XM_011511436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983464 c.3830C>G p.S1277C Chr.2: 127706504-127706504 on Build GRCh38
5774098 c.3796G>C p.G1266R Chr.2: 127706538-127706538 on Build GRCh38
1399284 c.3791delG p.G1264fs*17 Chr.2: 127706543-127706543 on Build GRCh38
1399283 c.3844G>A p.G1282R Chr.2: 127706490-127706490 on Build GRCh38
6821751 c.3840A>G p.L1280L Chr.2: 127706494-127706494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190867601 Chr.2: 127706640-127706640 on Build GRCh38
rs747684123 Chr.2: 127706530-127706530 on Build GRCh38
rs776151565 Chr.2: 127706581-127706582 on Build GRCh38
rs774334423 Chr.2: 127706548-127706548 on Build GRCh38
rs769356002 Chr.2: 127706531-127706531 on Build GRCh38
rs981803773 Chr.2: 127706678-127706678 on Build GRCh38
rs759799035 Chr.2: 127706549-127706549 on Build GRCh38
rs754304533 Chr.2: 127706586-127706586 on Build GRCh38
rs775625692 Chr.2: 127706569-127706569 on Build GRCh38
rs954157975 Chr.2: 127706484-127706484 on Build GRCh38
rs541597107 Chr.2: 127706545-127706545 on Build GRCh38
rs1019585748 Chr.2: 127706504-127706504 on Build GRCh38
rs764238608 Chr.2: 127706574-127706574 on Build GRCh38
rs544132979 Chr.2: 127706686-127706686 on Build GRCh38
rs779282664 Chr.2: 127706507-127706507 on Build GRCh38
rs771656881 Chr.2: 127706560-127706563 on Build GRCh38
rs369431202 Chr.2: 127706499-127706499 on Build GRCh38
rs1048845671 Chr.2: 127706652-127706652 on Build GRCh38
rs950244881 Chr.2: 127706602-127706602 on Build GRCh38
rs750829354 Chr.2: 127706598-127706598 on Build GRCh38
rs767698329 Chr.2: 127706563-127706563 on Build GRCh38
rs865856341 Chr.2: 127706506-127706506 on Build GRCh38
rs1044596694 Chr.2: 127706616-127706616 on Build GRCh38
rs764567706 Chr.2: 127706490-127706490 on Build GRCh38
rs765565621 Chr.2: 127706592-127706592 on Build GRCh38
rs754357420 Chr.2: 127706491-127706491 on Build GRCh38
rs564540606 Chr.2: 127706637-127706637 on Build GRCh38
rs34550770 Chr.2: 127706542-127706542 on Build GRCh38
rs757786255 Chr.2: 127706591-127706591 on Build GRCh38
rs749265737 Chr.2: 127706543-127706543 on Build GRCh38
rs780725829 Chr.2: 127706524-127706524 on Build GRCh38
rs970850419 Chr.2: 127706663-127706663 on Build GRCh38
rs754638633 Chr.2: 127706523-127706523 on Build GRCh38
rs760614243 Chr.2: 127706573-127706573 on Build GRCh38

View Full Product Details