Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154830644-154830917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM9
Entrez Gene ID
100132963
Gene Name
small integral membrane protein 9
Gene Aliases
CXorf68
UniGene
Hs.220762
Gene Chromosome Location
Chr.X: 154823348-154834662 on Build GRCh38
Transcripts
NM_001162936.1

SNP Detail

SNP ID Chromosome Location
rs886873382 Chr.X: 154830712-154830712 on Build GRCh38
rs782443981 Chr.X: 154830818-154830818 on Build GRCh38
rs376427110 Chr.X: 154830864-154830864 on Build GRCh38
rs782048065 Chr.X: 154830761-154830761 on Build GRCh38
rs12009271 Chr.X: 154830834-154830834 on Build GRCh38
rs184141613 Chr.X: 154830770-154830770 on Build GRCh38
rs940159688 Chr.X: 154830717-154830717 on Build GRCh38
rs199975971 Chr.X: 154830724-154830724 on Build GRCh38
rs782266601 Chr.X: 154830861-154830861 on Build GRCh38
rs782518347 Chr.X: 154830688-154830688 on Build GRCh38
rs891112446 Chr.X: 154830853-154830853 on Build GRCh38
rs192303481 Chr.X: 154830732-154830732 on Build GRCh38

View Full Product Details