Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114186865-114187136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB16
Entrez Gene ID
7704
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF, ZNF145
UniGene
Hs.591945
Gene Chromosome Location
Chr.11: 114059576-114256765 on Build GRCh38
Transcripts
NM_001018011.1 NM_006006.4 XM_005271658.4 XM_017018257.1 XM_017018258.1 XM_017018259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824992 c.1446C>A p.T482T Chr.11: 114187031-114187031 on Build GRCh38
4017985 c.1433C>G p.T478R Chr.11: 114187018-114187018 on Build GRCh38
1351412 c.1367C>T p.A456V Chr.11: 114186952-114186952 on Build GRCh38
5573395 c.1453+1G>T p.? Chr.11: 114187039-114187039 on Build GRCh38
6552686 c.1453G>T p.G485C Chr.11: 114187038-114187038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003170892 Chr.11: 114186991-114186991 on Build GRCh38
rs572169769 Chr.11: 114187084-114187084 on Build GRCh38
rs778386572 Chr.11: 114187065-114187065 on Build GRCh38
rs775275983 Chr.11: 114187088-114187088 on Build GRCh38
rs147151121 Chr.11: 114186964-114186964 on Build GRCh38
rs202099154 Chr.11: 114186998-114186998 on Build GRCh38
rs567057 Chr.11: 114186908-114186908 on Build GRCh38
rs765197054 Chr.11: 114187109-114187109 on Build GRCh38
rs758272725 Chr.11: 114187102-114187105 on Build GRCh38
rs776140047 Chr.11: 114187096-114187096 on Build GRCh38
rs770948069 Chr.11: 114186975-114186975 on Build GRCh38
rs761753131 Chr.11: 114186997-114186997 on Build GRCh38
rs781426011 Chr.11: 114187034-114187034 on Build GRCh38
rs1023160679 Chr.11: 114187097-114187097 on Build GRCh38
rs760244582 Chr.11: 114187092-114187092 on Build GRCh38
rs544966997 Chr.11: 114186928-114186928 on Build GRCh38
rs748661885 Chr.11: 114186938-114186938 on Build GRCh38
rs751718561 Chr.11: 114187027-114187027 on Build GRCh38
rs749709384 Chr.11: 114187075-114187075 on Build GRCh38
rs755153784 Chr.11: 114187031-114187031 on Build GRCh38
rs763364502 Chr.11: 114186953-114186953 on Build GRCh38
rs747154780 Chr.11: 114186922-114186922 on Build GRCh38
rs773729071 Chr.11: 114186949-114186949 on Build GRCh38
rs182657100 Chr.11: 114186901-114186901 on Build GRCh38
rs750339832 Chr.11: 114187111-114187111 on Build GRCh38
rs753275180 Chr.11: 114186984-114186984 on Build GRCh38
rs1010644921 Chr.11: 114186970-114186970 on Build GRCh38
rs768598164 Chr.11: 114186924-114186924 on Build GRCh38
rs116547293 Chr.11: 114186983-114186983 on Build GRCh38
rs570127168 Chr.11: 114187101-114187101 on Build GRCh38
rs542471640 Chr.11: 114186895-114186895 on Build GRCh38
rs758095993 Chr.11: 114187011-114187011 on Build GRCh38
rs760288157 Chr.11: 114186969-114186969 on Build GRCh38
rs771183852 Chr.11: 114187077-114187077 on Build GRCh38
rs372577096 Chr.11: 114187062-114187062 on Build GRCh38
rs780374276 Chr.11: 114187018-114187018 on Build GRCh38
rs748185557 Chr.11: 114187059-114187059 on Build GRCh38
rs530317678 Chr.11: 114186916-114186916 on Build GRCh38
rs771854572 Chr.11: 114186892-114186892 on Build GRCh38
rs770362959 Chr.11: 114186945-114186945 on Build GRCh38
rs972849524 Chr.11: 114187104-114187104 on Build GRCh38
rs998863957 Chr.11: 114187076-114187076 on Build GRCh38
rs750247832 Chr.11: 114187006-114187006 on Build GRCh38
rs768239185 Chr.11: 114187095-114187095 on Build GRCh38
rs541648463 Chr.11: 114187103-114187103 on Build GRCh38

View Full Product Details