Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50058954-50059185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC1
Entrez Gene ID
41
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2, ASIC, BNaC2
UniGene
Hs.274361
Gene Chromosome Location
Chr.12: 50057637-50083622 on Build GRCh38
Transcripts
NM_001095.3 NM_020039.3 NR_046389.1 XM_011538350.1 XM_011538351.2 XM_011538352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3954862 c.203A>C p.Y68S Chr.12: 50058969-50058969 on Build GRCh38
1705576 c.311C>A p.S104Y Chr.12: 50059077-50059077 on Build GRCh38
940286 c.315G>T p.K105N Chr.12: 50059081-50059081 on Build GRCh38
6605734 c.202T>C p.Y68H Chr.12: 50058968-50058968 on Build GRCh38
5361700 c.232G>A p.D78N Chr.12: 50058998-50058998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779174289 Chr.12: 50059145-50059145 on Build GRCh38
rs754000632 Chr.12: 50059139-50059139 on Build GRCh38
rs780194390 Chr.12: 50058998-50058998 on Build GRCh38
rs764985433 Chr.12: 50059098-50059098 on Build GRCh38
rs775314063 Chr.12: 50058979-50058979 on Build GRCh38
rs767328195 Chr.12: 50059135-50059135 on Build GRCh38
rs77090670 Chr.12: 50059148-50059148 on Build GRCh38
rs141895319 Chr.12: 50059069-50059069 on Build GRCh38
rs775264675 Chr.12: 50059103-50059103 on Build GRCh38
rs997398132 Chr.12: 50059126-50059126 on Build GRCh38
rs143760934 Chr.12: 50058997-50058997 on Build GRCh38
rs747530628 Chr.12: 50059062-50059062 on Build GRCh38
rs923136601 Chr.12: 50059146-50059146 on Build GRCh38
rs796514665 Chr.12: 50059102-50059102 on Build GRCh38
rs763984607 Chr.12: 50059113-50059113 on Build GRCh38
rs754999533 Chr.12: 50059143-50059143 on Build GRCh38
rs749289094 Chr.12: 50059001-50059001 on Build GRCh38
rs777915155 Chr.12: 50058984-50058984 on Build GRCh38
rs762907653 Chr.12: 50059104-50059104 on Build GRCh38
rs758935606 Chr.12: 50059093-50059093 on Build GRCh38
rs768965480 Chr.12: 50059003-50059003 on Build GRCh38
rs1052153922 Chr.12: 50059090-50059090 on Build GRCh38
rs924916569 Chr.12: 50059042-50059042 on Build GRCh38
rs779085870 Chr.12: 50059007-50059007 on Build GRCh38
rs370579463 Chr.12: 50059151-50059151 on Build GRCh38
rs776232671 Chr.12: 50059077-50059077 on Build GRCh38
rs773503565 Chr.12: 50059055-50059055 on Build GRCh38
rs1050666840 Chr.12: 50059127-50059127 on Build GRCh38
rs757239672 Chr.12: 50059123-50059123 on Build GRCh38
rs748571094 Chr.12: 50059039-50059039 on Build GRCh38
rs148123804 Chr.12: 50059054-50059054 on Build GRCh38
rs376321850 Chr.12: 50059136-50059136 on Build GRCh38
rs369668932 Chr.12: 50059149-50059149 on Build GRCh38
rs751340096 Chr.12: 50059117-50059117 on Build GRCh38
rs1043426733 Chr.12: 50059024-50059024 on Build GRCh38

View Full Product Details