Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62735333-62735602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC9C
Entrez Gene ID
283237
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
-
UniGene
Hs.31704
Gene Chromosome Location
Chr.11: 62728073-62738636 on Build GRCh38
Transcripts
NM_001318812.1 NM_001318813.1 NM_001318814.1 NM_001318815.1 NM_001318816.1 NM_173810.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5909189 c.239-1G>A p.? Chr.11: 62735381-62735381 on Build GRCh38
5970254 c.328G>A p.A110T Chr.11: 62735471-62735471 on Build GRCh38
2039581 c.349G>T p.A117S Chr.11: 62735492-62735492 on Build GRCh38
6069840 c.316G>C p.D106H Chr.11: 62735459-62735459 on Build GRCh38
6069841 c.279G>T p.V93V Chr.11: 62735422-62735422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765846607 Chr.11: 62735545-62735545 on Build GRCh38
rs753520382 Chr.11: 62735456-62735456 on Build GRCh38
rs376792542 Chr.11: 62735368-62735368 on Build GRCh38
rs376487549 Chr.11: 62735548-62735548 on Build GRCh38
rs547664703 Chr.11: 62735478-62735478 on Build GRCh38
rs117790598 Chr.11: 62735413-62735413 on Build GRCh38
rs755679896 Chr.11: 62735399-62735399 on Build GRCh38
rs748771281 Chr.11: 62735362-62735363 on Build GRCh38
rs778887758 Chr.11: 62735414-62735414 on Build GRCh38
rs977217538 Chr.11: 62735466-62735466 on Build GRCh38
rs1054335751 Chr.11: 62735507-62735507 on Build GRCh38
rs766002029 Chr.11: 62735454-62735454 on Build GRCh38
rs776891871 Chr.11: 62735440-62735440 on Build GRCh38
rs779594814 Chr.11: 62735356-62735357 on Build GRCh38
rs140788036 Chr.11: 62735447-62735447 on Build GRCh38
rs752652200 Chr.11: 62735480-62735480 on Build GRCh38
rs893779022 Chr.11: 62735362-62735362 on Build GRCh38
rs200921253 Chr.11: 62735513-62735513 on Build GRCh38
rs776822801 Chr.11: 62735358-62735358 on Build GRCh38
rs138276343 Chr.11: 62735492-62735492 on Build GRCh38
rs772634075 Chr.11: 62735451-62735451 on Build GRCh38
rs771098274 Chr.11: 62735435-62735435 on Build GRCh38
rs201691202 Chr.11: 62735502-62735502 on Build GRCh38
rs983282637 Chr.11: 62735517-62735517 on Build GRCh38
rs765894572 Chr.11: 62735374-62735374 on Build GRCh38
rs370398077 Chr.11: 62735372-62735372 on Build GRCh38
rs772603053 Chr.11: 62735387-62735388 on Build GRCh38
rs1009371476 Chr.11: 62735559-62735559 on Build GRCh38
rs779723199 Chr.11: 62735404-62735404 on Build GRCh38
rs769332416 Chr.11: 62735526-62735526 on Build GRCh38
rs539570547 Chr.11: 62735418-62735418 on Build GRCh38
rs931961512 Chr.11: 62735541-62735541 on Build GRCh38
rs774905674 Chr.11: 62735536-62735536 on Build GRCh38
rs1013524185 Chr.11: 62735573-62735573 on Build GRCh38
rs370622226 Chr.11: 62735377-62735377 on Build GRCh38
rs947060136 Chr.11: 62735482-62735482 on Build GRCh38
rs199784654 Chr.11: 62735385-62735385 on Build GRCh38
rs137950889 Chr.11: 62735445-62735445 on Build GRCh38
rs754771021 Chr.11: 62735471-62735471 on Build GRCh38
rs765047579 Chr.11: 62735561-62735561 on Build GRCh38
rs150104999 Chr.11: 62735450-62735450 on Build GRCh38
rs748011290 Chr.11: 62735417-62735417 on Build GRCh38
rs758293320 Chr.11: 62735481-62735481 on Build GRCh38
rs750107541 Chr.11: 62735395-62735395 on Build GRCh38
rs1043150410 Chr.11: 62735486-62735486 on Build GRCh38
rs767071798 Chr.11: 62735381-62735381 on Build GRCh38
rs781339476 Chr.11: 62735495-62735495 on Build GRCh38
rs775951621 Chr.11: 62735522-62735522 on Build GRCh38
rs556324819 Chr.11: 62735546-62735546 on Build GRCh38
rs370386237 Chr.11: 62735446-62735446 on Build GRCh38
rs201165764 Chr.11: 62735426-62735426 on Build GRCh38
rs753620423 Chr.11: 62735405-62735405 on Build GRCh38
rs749635226 Chr.11: 62735525-62735525 on Build GRCh38
rs747287467 Chr.11: 62735434-62735434 on Build GRCh38
rs747109982 Chr.11: 62735490-62735490 on Build GRCh38
rs752529356 Chr.11: 62735569-62735569 on Build GRCh38
rs777735013 Chr.11: 62735485-62735485 on Build GRCh38
rs760166766 Chr.11: 62735540-62735540 on Build GRCh38
rs57637308 Chr.11: 62735500-62735500 on Build GRCh38

View Full Product Details