Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24205933-24206182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
UniGene
Hs.314432
Gene Chromosome Location
Chr.14: 24205524-24208362 on Build GRCh38
Transcripts
NM_001184739.1 NM_001308067.1 NM_174944.3 XM_011536663.2 XM_017021226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
277862 c.154G>A p.A52T Chr.14: 24206077-24206077 on Build GRCh38
5419596 c.68A>G p.Y23C Chr.14: 24205991-24205991 on Build GRCh38
5040852 c.150G>C p.M50I Chr.14: 24206073-24206073 on Build GRCh38
5003955 c.38C>A p.T13N Chr.14: 24205961-24205961 on Build GRCh38
5419597 c.68A>G p.Y23C Chr.14: 24205991-24205991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778228421 Chr.14: 24206135-24206135 on Build GRCh38
rs1055884921 Chr.14: 24206014-24206015 on Build GRCh38
rs765213244 Chr.14: 24206007-24206008 on Build GRCh38
rs768889685 Chr.14: 24206133-24206133 on Build GRCh38
rs756680087 Chr.14: 24206068-24206068 on Build GRCh38
rs777488097 Chr.14: 24205976-24205976 on Build GRCh38
rs762807506 Chr.14: 24206095-24206097 on Build GRCh38
rs747352590 Chr.14: 24206126-24206126 on Build GRCh38
rs776821940 Chr.14: 24206025-24206025 on Build GRCh38
rs772554013 Chr.14: 24206103-24206103 on Build GRCh38
rs36036137 Chr.14: 24206020-24206020 on Build GRCh38
rs761643462 Chr.14: 24205983-24205983 on Build GRCh38
rs886841369 Chr.14: 24206048-24206048 on Build GRCh38
rs764270231 Chr.14: 24206017-24206017 on Build GRCh38
rs755867204 Chr.14: 24206052-24206052 on Build GRCh38
rs769561757 Chr.14: 24205970-24205970 on Build GRCh38
rs146066616 Chr.14: 24206027-24206027 on Build GRCh38
rs752531838 Chr.14: 24206041-24206041 on Build GRCh38
rs775874092 Chr.14: 24206115-24206115 on Build GRCh38
rs996483726 Chr.14: 24206030-24206030 on Build GRCh38
rs376850456 Chr.14: 24206002-24206002 on Build GRCh38
rs748850454 Chr.14: 24206065-24206065 on Build GRCh38
rs748712620 Chr.14: 24205993-24205993 on Build GRCh38
rs772354818 Chr.14: 24206009-24206009 on Build GRCh38
rs751388870 Chr.14: 24206033-24206033 on Build GRCh38
rs776679657 Chr.14: 24206140-24206140 on Build GRCh38
rs963949076 Chr.14: 24206149-24206149 on Build GRCh38
rs765886696 Chr.14: 24206157-24206157 on Build GRCh38
rs779477458 Chr.14: 24206069-24206069 on Build GRCh38
rs760954275 Chr.14: 24206015-24206015 on Build GRCh38
rs375430080 Chr.14: 24206075-24206077 on Build GRCh38
rs141363891 Chr.14: 24206158-24206158 on Build GRCh38
rs371150247 Chr.14: 24206005-24206005 on Build GRCh38
rs746302677 Chr.14: 24206097-24206097 on Build GRCh38
rs746273198 Chr.14: 24206008-24206008 on Build GRCh38
rs960183312 Chr.14: 24206138-24206138 on Build GRCh38
rs756478429 Chr.14: 24206037-24206037 on Build GRCh38
rs750353923 Chr.14: 24206063-24206065 on Build GRCh38
rs1008179126 Chr.14: 24205979-24205979 on Build GRCh38
rs201368541 Chr.14: 24206036-24206036 on Build GRCh38
rs1015737252 Chr.14: 24206137-24206137 on Build GRCh38
rs761960213 Chr.14: 24206156-24206156 on Build GRCh38
rs775702618 Chr.14: 24206010-24206010 on Build GRCh38
rs777543019 Chr.14: 24206063-24206063 on Build GRCh38
rs201948249 Chr.14: 24206112-24206112 on Build GRCh38

View Full Product Details