Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23568142-23568367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST9L
Entrez Gene ID
128821
Gene Name
cystatin 9-like
Gene Aliases
CTES7B, bA218C14.1
UniGene
Hs.121554
Gene Chromosome Location
Chr.20: 23564732-23568749 on Build GRCh38
Transcripts
NM_080610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4939624 c.229T>C p.W77R Chr.20: 23568222-23568222 on Build GRCh38
3991583 c.219C>A p.I73I Chr.20: 23568232-23568232 on Build GRCh38
1326846 c.199G>T p.A67S Chr.20: 23568252-23568252 on Build GRCh38
4336725 c.213_214insG p.H72fs*26 Chr.20: 23568237-23568238 on Build GRCh38
4097163 c.211G>T p.G71W Chr.20: 23568240-23568240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775930017 Chr.20: 23568316-23568316 on Build GRCh38
rs975696919 Chr.20: 23568345-23568345 on Build GRCh38
rs113667569 Chr.20: 23568282-23568282 on Build GRCh38
rs200065971 Chr.20: 23568281-23568281 on Build GRCh38
rs746690420 Chr.20: 23568314-23568314 on Build GRCh38
rs895130545 Chr.20: 23568195-23568195 on Build GRCh38
rs775815886 Chr.20: 23568232-23568232 on Build GRCh38
rs61743019 Chr.20: 23568266-23568266 on Build GRCh38
rs141989904 Chr.20: 23568290-23568290 on Build GRCh38
rs371284976 Chr.20: 23568196-23568196 on Build GRCh38
rs558085521 Chr.20: 23568310-23568310 on Build GRCh38
rs771771836 Chr.20: 23568210-23568210 on Build GRCh38
rs769999853 Chr.20: 23568315-23568315 on Build GRCh38
rs774548597 Chr.20: 23568318-23568318 on Build GRCh38
rs747707920 Chr.20: 23568308-23568308 on Build GRCh38
rs200161050 Chr.20: 23568287-23568287 on Build GRCh38
rs746271617 Chr.20: 23568216-23568216 on Build GRCh38
rs757560305 Chr.20: 23568181-23568181 on Build GRCh38
rs762905249 Chr.20: 23568172-23568172 on Build GRCh38
rs989426002 Chr.20: 23568329-23568329 on Build GRCh38
rs570491182 Chr.20: 23568183-23568183 on Build GRCh38
rs767733896 Chr.20: 23568331-23568331 on Build GRCh38
rs778382805 Chr.20: 23568303-23568303 on Build GRCh38
rs374586618 Chr.20: 23568173-23568173 on Build GRCh38
rs371800048 Chr.20: 23568274-23568274 on Build GRCh38
rs762020452 Chr.20: 23568320-23568320 on Build GRCh38
rs773518554 Chr.20: 23568338-23568338 on Build GRCh38
rs749278295 Chr.20: 23568198-23568198 on Build GRCh38
rs758015425 Chr.20: 23568309-23568309 on Build GRCh38
rs760965696 Chr.20: 23568342-23568342 on Build GRCh38
rs763533371 Chr.20: 23568238-23568238 on Build GRCh38
rs201702074 Chr.20: 23568311-23568311 on Build GRCh38
rs62207002 Chr.20: 23568199-23568199 on Build GRCh38
rs200746390 Chr.20: 23568182-23568182 on Build GRCh38
rs926061051 Chr.20: 23568283-23568283 on Build GRCh38
rs535740829 Chr.20: 23568211-23568211 on Build GRCh38
rs752792336 Chr.20: 23568163-23568163 on Build GRCh38
rs774642821 Chr.20: 23568250-23568250 on Build GRCh38
rs200398348 Chr.20: 23568317-23568317 on Build GRCh38
rs578026236 Chr.20: 23568347-23568347 on Build GRCh38
rs376346940 Chr.20: 23568280-23568280 on Build GRCh38
rs770225965 Chr.20: 23568229-23568229 on Build GRCh38
rs369382363 Chr.20: 23568206-23568206 on Build GRCh38
rs751776236 Chr.20: 23568178-23568178 on Build GRCh38
rs200554161 Chr.20: 23568202-23568202 on Build GRCh38
rs756094976 Chr.20: 23568193-23568193 on Build GRCh38

View Full Product Details