Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 97981028-97981289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STPG2
Entrez Gene ID
285555
Gene Name
sperm tail PG-rich repeat containing 2
Gene Aliases
C4orf37
UniGene
Hs.680226
Gene Chromosome Location
Chr.4: 97441450-98143473 on Build GRCh38
Transcripts
NM_174952.2 XM_011531886.2 XM_017008049.1 XM_017008050.1 XM_017008051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
199267 c.763G>A p.E255K Chr.4: 97981168-97981168 on Build GRCh38
1198852 c.757A>C p.T253P Chr.4: 97981174-97981174 on Build GRCh38
4918304 c.659A>T p.N220I Chr.4: 97981272-97981272 on Build GRCh38
7142926 c.736C>A p.R246R Chr.4: 97981195-97981195 on Build GRCh38
5679740 c.667C>T p.R223* Chr.4: 97981264-97981264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150708038 Chr.4: 97981263-97981263 on Build GRCh38
rs963494322 Chr.4: 97981087-97981087 on Build GRCh38
rs201834612 Chr.4: 97981264-97981264 on Build GRCh38
rs759785509 Chr.4: 97981198-97981198 on Build GRCh38
rs142056067 Chr.4: 97981259-97981259 on Build GRCh38
rs973569094 Chr.4: 97981107-97981107 on Build GRCh38
rs775780937 Chr.4: 97981235-97981235 on Build GRCh38
rs913275838 Chr.4: 97981237-97981237 on Build GRCh38
rs1039329550 Chr.4: 97981097-97981097 on Build GRCh38
rs754300557 Chr.4: 97981217-97981217 on Build GRCh38
rs567995463 Chr.4: 97981061-97981061 on Build GRCh38
rs763948719 Chr.4: 97981126-97981126 on Build GRCh38
rs750242856 Chr.4: 97981258-97981258 on Build GRCh38
rs547460502 Chr.4: 97981253-97981253 on Build GRCh38
rs199854534 Chr.4: 97981216-97981216 on Build GRCh38
rs1057220396 Chr.4: 97981189-97981189 on Build GRCh38
rs755770186 Chr.4: 97981141-97981141 on Build GRCh38
rs776903037 Chr.4: 97981196-97981196 on Build GRCh38
rs35146453 Chr.4: 97981127-97981127 on Build GRCh38
rs764605064 Chr.4: 97981190-97981190 on Build GRCh38
rs960152021 Chr.4: 97981207-97981207 on Build GRCh38
rs758323878 Chr.4: 97981123-97981123 on Build GRCh38
rs757224302 Chr.4: 97981255-97981255 on Build GRCh38
rs267600307 Chr.4: 97981168-97981168 on Build GRCh38
rs780878638 Chr.4: 97981136-97981136 on Build GRCh38
rs779602150 Chr.4: 97981151-97981151 on Build GRCh38
rs567902903 Chr.4: 97981206-97981206 on Build GRCh38
rs761678606 Chr.4: 97981109-97981109 on Build GRCh38
rs1001565083 Chr.4: 97981202-97981202 on Build GRCh38
rs762959299 Chr.4: 97981238-97981238 on Build GRCh38
rs375848968 Chr.4: 97981195-97981195 on Build GRCh38
rs900952264 Chr.4: 97981119-97981119 on Build GRCh38
rs773798698 Chr.4: 97981190-97981190 on Build GRCh38
rs112678572 Chr.4: 97981233-97981233 on Build GRCh38
rs76740552 Chr.4: 97981252-97981252 on Build GRCh38
rs745500755 Chr.4: 97981140-97981140 on Build GRCh38
rs756409350 Chr.4: 97981133-97981138 on Build GRCh38
rs868022016 Chr.4: 97981161-97981161 on Build GRCh38
rs772987445 Chr.4: 97981149-97981149 on Build GRCh38
rs922265239 Chr.4: 97981266-97981266 on Build GRCh38
rs370875603 Chr.4: 97981174-97981174 on Build GRCh38
rs192279668 Chr.4: 97981073-97981073 on Build GRCh38
rs117860793 Chr.4: 97981256-97981256 on Build GRCh38
rs113380860 Chr.4: 97981254-97981254 on Build GRCh38
rs748816795 Chr.4: 97981153-97981153 on Build GRCh38
rs201860057 Chr.4: 97981194-97981194 on Build GRCh38
rs768286575 Chr.4: 97981157-97981157 on Build GRCh38
rs756995338 Chr.4: 97981130-97981130 on Build GRCh38

View Full Product Details