Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54378103-54378364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709347 c.1110G>A p.P370P Chr.18: 54378237-54378237 on Build GRCh38
4072598 c.1102C>T p.R368W Chr.18: 54378229-54378229 on Build GRCh38
2879993 c.1092G>A p.T364T Chr.18: 54378219-54378219 on Build GRCh38
6614812 c.1091C>T p.T364M Chr.18: 54378218-54378218 on Build GRCh38
3526478 c.1077C>T p.D359D Chr.18: 54378204-54378204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530382764 Chr.18: 54378270-54378270 on Build GRCh38
rs764748945 Chr.18: 54378175-54378175 on Build GRCh38
rs775707183 Chr.18: 54378166-54378166 on Build GRCh38
rs551048507 Chr.18: 54378202-54378202 on Build GRCh38
rs561960087 Chr.18: 54378219-54378219 on Build GRCh38
rs1041980323 Chr.18: 54378214-54378214 on Build GRCh38
rs758741714 Chr.18: 54378275-54378275 on Build GRCh38
rs759820185 Chr.18: 54378162-54378162 on Build GRCh38
rs375717506 Chr.18: 54378168-54378168 on Build GRCh38
rs769696402 Chr.18: 54378165-54378165 on Build GRCh38
rs372860791 Chr.18: 54378271-54378274 on Build GRCh38
rs3753057 Chr.18: 54378128-54378128 on Build GRCh38
rs573009456 Chr.18: 54378148-54378148 on Build GRCh38
rs958025437 Chr.18: 54378286-54378286 on Build GRCh38
rs546816755 Chr.18: 54378309-54378309 on Build GRCh38
rs765791921 Chr.18: 54378205-54378205 on Build GRCh38
rs753436104 Chr.18: 54378206-54378206 on Build GRCh38
rs755030286 Chr.18: 54378148-54378148 on Build GRCh38
rs751945039 Chr.18: 54378176-54378176 on Build GRCh38
rs577158137 Chr.18: 54378228-54378228 on Build GRCh38
rs371938513 Chr.18: 54378138-54378138 on Build GRCh38
rs1037419516 Chr.18: 54378245-54378245 on Build GRCh38
rs865944318 Chr.18: 54378300-54378300 on Build GRCh38
rs377117634 Chr.18: 54378265-54378265 on Build GRCh38
rs749974011 Chr.18: 54378193-54378193 on Build GRCh38
rs941774615 Chr.18: 54378243-54378243 on Build GRCh38
rs141811954 Chr.18: 54378177-54378177 on Build GRCh38
rs749430389 Chr.18: 54378237-54378237 on Build GRCh38
rs778781471 Chr.18: 54378218-54378218 on Build GRCh38
rs886079327 Chr.18: 54378236-54378236 on Build GRCh38
rs943013534 Chr.18: 54378320-54378320 on Build GRCh38
rs766806300 Chr.18: 54378196-54378196 on Build GRCh38
rs942273565 Chr.18: 54378250-54378250 on Build GRCh38
rs752896443 Chr.18: 54378271-54378273 on Build GRCh38
rs916152681 Chr.18: 54378235-54378235 on Build GRCh38
rs3753055 Chr.18: 54378274-54378274 on Build GRCh38
rs1051947755 Chr.18: 54378275-54378275 on Build GRCh38
rs3753056 Chr.18: 54378271-54378271 on Build GRCh38
rs938584674 Chr.18: 54378230-54378230 on Build GRCh38
rs1023756004 Chr.18: 54378334-54378334 on Build GRCh38
rs558955707 Chr.18: 54378183-54378183 on Build GRCh38
rs777550164 Chr.18: 54378223-54378223 on Build GRCh38
rs137873678 Chr.18: 54378211-54378211 on Build GRCh38
rs148065410 Chr.18: 54378233-54378233 on Build GRCh38
rs1035814700 Chr.18: 54378221-54378221 on Build GRCh38
rs146934571 Chr.18: 54378194-54378194 on Build GRCh38
rs544918205 Chr.18: 54378229-54378229 on Build GRCh38

View Full Product Details