Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92141056-92141281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
-
UniGene
Hs.676102
Gene Chromosome Location
Chr.1: 92080305-92147840 on Build GRCh38
Transcripts
NM_183242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5643328 c.905G>A p.R302K Chr.1: 92141186-92141186 on Build GRCh38
3866244 c.925C>T p.Q309* Chr.1: 92141206-92141206 on Build GRCh38
5643327 c.902G>T p.R301I Chr.1: 92141183-92141183 on Build GRCh38
3966807 c.871G>T p.G291* Chr.1: 92141152-92141152 on Build GRCh38
913264 c.921C>T p.F307F Chr.1: 92141202-92141202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764581109 Chr.1: 92141125-92141125 on Build GRCh38
rs771933710 Chr.1: 92141101-92141101 on Build GRCh38
rs377434732 Chr.1: 92141163-92141163 on Build GRCh38
rs778630377 Chr.1: 92141089-92141089 on Build GRCh38
rs773202811 Chr.1: 92141115-92141115 on Build GRCh38
rs776789074 Chr.1: 92141105-92141105 on Build GRCh38
rs765893275 Chr.1: 92141162-92141162 on Build GRCh38
rs763215425 Chr.1: 92141119-92141119 on Build GRCh38
rs200581113 Chr.1: 92141242-92141242 on Build GRCh38
rs769915151 Chr.1: 92141113-92141113 on Build GRCh38
rs778685281 Chr.1: 92141185-92141185 on Build GRCh38
rs746443590 Chr.1: 92141092-92141092 on Build GRCh38
rs758203077 Chr.1: 92141202-92141202 on Build GRCh38
rs939452150 Chr.1: 92141148-92141148 on Build GRCh38
rs372506597 Chr.1: 92141126-92141126 on Build GRCh38
rs761648991 Chr.1: 92141109-92141110 on Build GRCh38
rs762935549 Chr.1: 92141121-92141124 on Build GRCh38
rs776220827 Chr.1: 92141099-92141099 on Build GRCh38
rs775621769 Chr.1: 92141118-92141118 on Build GRCh38
rs769548697 Chr.1: 92141111-92141112 on Build GRCh38
rs755618646 Chr.1: 92141138-92141138 on Build GRCh38
rs767966681 Chr.1: 92141131-92141131 on Build GRCh38
rs754585139 Chr.1: 92141180-92141180 on Build GRCh38
rs565396333 Chr.1: 92141095-92141095 on Build GRCh38
rs768354666 Chr.1: 92141094-92141095 on Build GRCh38
rs747883887 Chr.1: 92141191-92141191 on Build GRCh38
rs141428099 Chr.1: 92141106-92141106 on Build GRCh38
rs963899898 Chr.1: 92141235-92141235 on Build GRCh38
rs750935477 Chr.1: 92141135-92141135 on Build GRCh38
rs992397161 Chr.1: 92141164-92141164 on Build GRCh38
rs754817984 Chr.1: 92141087-92141087 on Build GRCh38
rs762423196 Chr.1: 92141127-92141127 on Build GRCh38

View Full Product Details