Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7655544-7655777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B2
Entrez Gene ID
482
Gene Name
ATPase Na+/K+ transporting subunit beta 2
Gene Aliases
AMOG
UniGene
Hs.643540
Gene Chromosome Location
Chr.17: 7646627-7657771 on Build GRCh38
Transcripts
NM_001303263.1 NM_001678.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139862 c.708+7C>A p.? Chr.17: 7655632-7655632 on Build GRCh38
84369 c.692A>T p.Y231F Chr.17: 7655609-7655609 on Build GRCh38
6326222 c.706C>T p.H236Y Chr.17: 7655623-7655623 on Build GRCh38
291955 c.726C>A p.P242P Chr.17: 7655748-7655748 on Build GRCh38
4166724 c.655G>A p.A219T Chr.17: 7655572-7655572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781608500 Chr.17: 7655751-7655751 on Build GRCh38
rs764745784 Chr.17: 7655586-7655586 on Build GRCh38
rs529731078 Chr.17: 7655587-7655587 on Build GRCh38
rs749738030 Chr.17: 7655748-7655748 on Build GRCh38
rs754744851 Chr.17: 7655606-7655606 on Build GRCh38
rs748581309 Chr.17: 7655725-7655725 on Build GRCh38
rs767443988 Chr.17: 7655689-7655689 on Build GRCh38
rs780151975 Chr.17: 7655661-7655661 on Build GRCh38
rs762432825 Chr.17: 7655681-7655681 on Build GRCh38
rs772222418 Chr.17: 7655568-7655568 on Build GRCh38
rs1031593909 Chr.17: 7655746-7655746 on Build GRCh38
rs761002067 Chr.17: 7655572-7655572 on Build GRCh38
rs780137295 Chr.17: 7655711-7655711 on Build GRCh38
rs375845891 Chr.17: 7655665-7655665 on Build GRCh38
rs770134722 Chr.17: 7655727-7655727 on Build GRCh38
rs765858689 Chr.17: 7655687-7655687 on Build GRCh38
rs781363639 Chr.17: 7655723-7655723 on Build GRCh38
rs189871284 Chr.17: 7655619-7655619 on Build GRCh38
rs150919258 Chr.17: 7655594-7655594 on Build GRCh38
rs559593930 Chr.17: 7655656-7655656 on Build GRCh38
rs113495677 Chr.17: 7655743-7655743 on Build GRCh38
rs770328897 Chr.17: 7655675-7655675 on Build GRCh38
rs202126873 Chr.17: 7655633-7655633 on Build GRCh38
rs371442690 Chr.17: 7655676-7655676 on Build GRCh38
rs773609058 Chr.17: 7655732-7655732 on Build GRCh38
rs762497585 Chr.17: 7655592-7655592 on Build GRCh38
rs1001012398 Chr.17: 7655724-7655724 on Build GRCh38
rs1049388743 Chr.17: 7655635-7655635 on Build GRCh38
rs574358752 Chr.17: 7655571-7655571 on Build GRCh38
rs55831773 Chr.17: 7655719-7655719 on Build GRCh38
rs200903956 Chr.17: 7655688-7655688 on Build GRCh38
rs755239918 Chr.17: 7655720-7655720 on Build GRCh38
rs762264628 Chr.17: 7655578-7655578 on Build GRCh38
rs35919584 Chr.17: 7655571-7655572 on Build GRCh38
rs751214466 Chr.17: 7655601-7655601 on Build GRCh38
rs368911633 Chr.17: 7655690-7655690 on Build GRCh38
rs762146423 Chr.17: 7655667-7655667 on Build GRCh38
rs781070046 Chr.17: 7655613-7655613 on Build GRCh38
rs954193078 Chr.17: 7655651-7655651 on Build GRCh38
rs777821069 Chr.17: 7655653-7655653 on Build GRCh38
rs758409378 Chr.17: 7655708-7655708 on Build GRCh38
rs117613045 Chr.17: 7655577-7655577 on Build GRCh38
rs376822561 Chr.17: 7655657-7655657 on Build GRCh38
rs371912048 Chr.17: 7655663-7655663 on Build GRCh38
rs200592951 Chr.17: 7655704-7655704 on Build GRCh38
rs769004384 Chr.17: 7655576-7655576 on Build GRCh38
rs367554240 Chr.17: 7655662-7655662 on Build GRCh38

View Full Product Details