Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41012627-41012852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B6
Entrez Gene ID
1555
Gene Name
cytochrome P450 family 2 subfamily B member 6
Gene Aliases
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6, EFVM, IIB1, P450
UniGene
Hs.1360
Gene Chromosome Location
Chr.19: 40991200-41018402 on Build GRCh38
Transcripts
NM_000767.4 XM_005258569.4 XM_006723050.3 XM_011526546.2 XM_011526547.2 XM_011526548.2 XM_011526549.2 XM_011526550.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534271 c.1210A>G p.K404E Chr.19: 41012731-41012731 on Build GRCh38
3534272 c.1281C>T p.I427I Chr.19: 41012802-41012802 on Build GRCh38
4894441 c.1188C>T p.L396L Chr.19: 41012709-41012709 on Build GRCh38
1721780 c.1189C>T p.H397Y Chr.19: 41012710-41012710 on Build GRCh38
6639175 c.1218C>T p.D406D Chr.19: 41012739-41012739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35979566 Chr.19: 41012693-41012693 on Build GRCh38
rs17411404 Chr.19: 41012789-41012789 on Build GRCh38
rs767042783 Chr.19: 41012651-41012651 on Build GRCh38
rs777953664 Chr.19: 41012757-41012757 on Build GRCh38
rs146634957 Chr.19: 41012799-41012799 on Build GRCh38
rs35449271 Chr.19: 41012668-41012668 on Build GRCh38
rs35010579 Chr.19: 41012822-41012822 on Build GRCh38
rs757061955 Chr.19: 41012671-41012671 on Build GRCh38
rs768220817 Chr.19: 41012808-41012810 on Build GRCh38
rs775031571 Chr.19: 41012774-41012774 on Build GRCh38
rs35661880 Chr.19: 41012739-41012739 on Build GRCh38
rs867514011 Chr.19: 41012719-41012719 on Build GRCh38
rs755094512 Chr.19: 41012829-41012829 on Build GRCh38
rs267605495 Chr.19: 41012815-41012815 on Build GRCh38
rs750385837 Chr.19: 41012679-41012679 on Build GRCh38
rs751743352 Chr.19: 41012828-41012828 on Build GRCh38
rs35010098 Chr.19: 41012803-41012803 on Build GRCh38
rs764884197 Chr.19: 41012675-41012675 on Build GRCh38
rs751655485 Chr.19: 41012697-41012697 on Build GRCh38
rs761312899 Chr.19: 41012802-41012802 on Build GRCh38
rs749827793 Chr.19: 41012758-41012758 on Build GRCh38
rs763770351 Chr.19: 41012665-41012665 on Build GRCh38
rs193922918 Chr.19: 41012740-41012740 on Build GRCh38
rs763021653 Chr.19: 41012813-41012813 on Build GRCh38
rs150072531 Chr.19: 41012711-41012711 on Build GRCh38
rs372874729 Chr.19: 41012776-41012776 on Build GRCh38
rs748783536 Chr.19: 41012772-41012773 on Build GRCh38
rs569717498 Chr.19: 41012704-41012704 on Build GRCh38
rs768260761 Chr.19: 41012795-41012795 on Build GRCh38
rs867126676 Chr.19: 41012773-41012773 on Build GRCh38
rs140993284 Chr.19: 41012674-41012674 on Build GRCh38
rs530338921 Chr.19: 41012689-41012689 on Build GRCh38
rs145114902 Chr.19: 41012811-41012811 on Build GRCh38
rs771398074 Chr.19: 41012771-41012771 on Build GRCh38
rs774929747 Chr.19: 41012653-41012653 on Build GRCh38
rs370464706 Chr.19: 41012659-41012659 on Build GRCh38

View Full Product Details