Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9528436-9528661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF426
Entrez Gene ID
79088
Gene Name
zinc finger protein 426
Gene Aliases
K-RBP
UniGene
Hs.386476
Gene Chromosome Location
Chr.19: 9523224-9538673 on Build GRCh38
Transcripts
NM_001300883.2 NM_001318055.1 NM_001318056.1 NM_024106.2 XM_017027293.1 XM_017027294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189859 c.1544G>C p.C515S Chr.19: 9528501-9528501 on Build GRCh38
6830136 c.1600T>A p.C534S Chr.19: 9528445-9528445 on Build GRCh38
6441804 c.1592C>T p.P531L Chr.19: 9528453-9528453 on Build GRCh38
4468803 c.1564C>T p.H522Y Chr.19: 9528481-9528481 on Build GRCh38
2702142 c.1541C>T p.T514M Chr.19: 9528504-9528504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150850267 Chr.19: 9528534-9528534 on Build GRCh38
rs749347897 Chr.19: 9528551-9528551 on Build GRCh38
rs868339840 Chr.19: 9528492-9528492 on Build GRCh38
rs1012539121 Chr.19: 9528516-9528516 on Build GRCh38
rs777154730 Chr.19: 9528617-9528618 on Build GRCh38
rs1025051211 Chr.19: 9528509-9528509 on Build GRCh38
rs1012190313 Chr.19: 9528505-9528505 on Build GRCh38
rs760269190 Chr.19: 9528635-9528635 on Build GRCh38
rs140022666 Chr.19: 9528636-9528636 on Build GRCh38
rs7351067 Chr.19: 9528593-9528593 on Build GRCh38
rs757187141 Chr.19: 9528512-9528512 on Build GRCh38
rs769934986 Chr.19: 9528470-9528470 on Build GRCh38
rs763506198 Chr.19: 9528473-9528473 on Build GRCh38
rs759835593 Chr.19: 9528588-9528588 on Build GRCh38
rs755628056 Chr.19: 9528501-9528501 on Build GRCh38
rs778874473 Chr.19: 9528515-9528515 on Build GRCh38
rs200145637 Chr.19: 9528471-9528471 on Build GRCh38
rs771408767 Chr.19: 9528563-9528563 on Build GRCh38
rs752292097 Chr.19: 9528497-9528497 on Build GRCh38
rs745716524 Chr.19: 9528518-9528518 on Build GRCh38
rs371400377 Chr.19: 9528475-9528475 on Build GRCh38
rs79497129 Chr.19: 9528503-9528503 on Build GRCh38
rs749740115 Chr.19: 9528619-9528620 on Build GRCh38
rs200716581 Chr.19: 9528504-9528504 on Build GRCh38
rs758198893 Chr.19: 9528530-9528530 on Build GRCh38
rs774911721 Chr.19: 9528570-9528570 on Build GRCh38
rs144384307 Chr.19: 9528538-9528538 on Build GRCh38
rs759944931 Chr.19: 9528476-9528476 on Build GRCh38
rs775158234 Chr.19: 9528627-9528627 on Build GRCh38
rs767136395 Chr.19: 9528480-9528480 on Build GRCh38
rs773477507 Chr.19: 9528546-9528546 on Build GRCh38
rs767857958 Chr.19: 9528616-9528616 on Build GRCh38
rs781576924 Chr.19: 9528531-9528531 on Build GRCh38

View Full Product Details