Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32704315-32704588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD7
Entrez Gene ID
149951
Gene Name
COMM domain containing 7
Gene Aliases
C20orf92, dJ1085F17.3
UniGene
Hs.408427
Gene Chromosome Location
Chr.20: 32702691-32744008 on Build GRCh38
Transcripts
NM_001099339.1 NM_053041.2 XM_005260299.3 XM_005260300.3 XM_011528604.2 XM_017027685.1 XM_017027686.1 XM_017027687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495033 c.448T>G p.L150V Chr.20: 32704469-32704469 on Build GRCh38
3749697 c.445G>A p.E149K Chr.20: 32704472-32704472 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs887962678 Chr.20: 32704514-32704514 on Build GRCh38
rs912861353 Chr.20: 32704375-32704375 on Build GRCh38
rs975784617 Chr.20: 32704409-32704409 on Build GRCh38
rs757803367 Chr.20: 32704451-32704451 on Build GRCh38
rs766763605 Chr.20: 32704526-32704527 on Build GRCh38
rs762939645 Chr.20: 32704510-32704512 on Build GRCh38
rs199774981 Chr.20: 32704510-32704510 on Build GRCh38
rs780253669 Chr.20: 32704496-32704496 on Build GRCh38
rs778100304 Chr.20: 32704497-32704500 on Build GRCh38
rs962486945 Chr.20: 32704535-32704535 on Build GRCh38
rs754358204 Chr.20: 32704496-32704496 on Build GRCh38
rs774392318 Chr.20: 32704518-32704518 on Build GRCh38
rs555257837 Chr.20: 32704473-32704473 on Build GRCh38
rs537646194 Chr.20: 32704566-32704566 on Build GRCh38
rs769605312 Chr.20: 32704508-32704514 on Build GRCh38
rs760303681 Chr.20: 32704511-32704516 on Build GRCh38
rs201349403 Chr.20: 32704497-32704498 on Build GRCh38
rs57986478 Chr.20: 32704384-32704385 on Build GRCh38
rs1050586644 Chr.20: 32704453-32704453 on Build GRCh38
rs750110988 Chr.20: 32704448-32704449 on Build GRCh38
rs768924004 Chr.20: 32704516-32704516 on Build GRCh38
rs753448295 Chr.20: 32704394-32704394 on Build GRCh38
rs142629794 Chr.20: 32704348-32704348 on Build GRCh38
rs747232593 Chr.20: 32704508-32704512 on Build GRCh38
rs771958080 Chr.20: 32704534-32704534 on Build GRCh38
rs59379723 Chr.20: 32704511-32704511 on Build GRCh38
rs774296487 Chr.20: 32704510-32704511 on Build GRCh38
rs775204335 Chr.20: 32704509-32704514 on Build GRCh38
rs138467153 Chr.20: 32704382-32704383 on Build GRCh38
rs945684561 Chr.20: 32704378-32704378 on Build GRCh38
rs879222338 Chr.20: 32704512-32704514 on Build GRCh38
rs370081454 Chr.20: 32704472-32704472 on Build GRCh38
rs768525735 Chr.20: 32704510-32704514 on Build GRCh38
rs72183305 Chr.20: 32704383-32704384 on Build GRCh38
rs930308073 Chr.20: 32704530-32704530 on Build GRCh38
rs957285433 Chr.20: 32704352-32704352 on Build GRCh38
rs756471449 Chr.20: 32704494-32704495 on Build GRCh38
rs972841262 Chr.20: 32704544-32704544 on Build GRCh38
rs748179593 Chr.20: 32704497-32704499 on Build GRCh38
rs749505820 Chr.20: 32704501-32704501 on Build GRCh38
rs777653329 Chr.20: 32704371-32704371 on Build GRCh38
rs772300786 Chr.20: 32704496-32704497 on Build GRCh38
rs573634023 Chr.20: 32704555-32704555 on Build GRCh38
rs767057208 Chr.20: 32704511-32704512 on Build GRCh38
rs111526861 Chr.20: 32704508-32704508 on Build GRCh38
rs748266791 Chr.20: 32704526-32704526 on Build GRCh38
rs781576753 Chr.20: 32704463-32704463 on Build GRCh38
rs780332590 Chr.20: 32704495-32704496 on Build GRCh38
rs767110274 Chr.20: 32704429-32704431 on Build GRCh38
rs377633951 Chr.20: 32704511-32704511 on Build GRCh38
rs750068657 Chr.20: 32704511-32704514 on Build GRCh38
rs754473770 Chr.20: 32704406-32704406 on Build GRCh38
rs200530368 Chr.20: 32704445-32704445 on Build GRCh38
rs377565040 Chr.20: 32704411-32704411 on Build GRCh38

View Full Product Details