Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23499661-23499886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2E2
Entrez Gene ID
7325
Gene Name
ubiquitin conjugating enzyme E2 E2
Gene Aliases
UBCH8
UniGene
Hs.475688
Gene Chromosome Location
Chr.3: 23203007-23590805 on Build GRCh38
Transcripts
NM_152653.3 XM_005265432.2 XM_005265433.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479988 c.314T>C p.F105S Chr.3: 23499694-23499694 on Build GRCh38
3427485 c.330T>G p.F110L Chr.3: 23499710-23499710 on Build GRCh38
5934839 c.316C>T p.L106F Chr.3: 23499696-23499696 on Build GRCh38
4738672 c.360G>T p.K120N Chr.3: 23499740-23499740 on Build GRCh38
4584234 c.285A>C p.P95P Chr.3: 23499665-23499665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949371617 Chr.3: 23499789-23499789 on Build GRCh38
rs746872939 Chr.3: 23499747-23499747 on Build GRCh38
rs1032470970 Chr.3: 23499846-23499846 on Build GRCh38
rs768382348 Chr.3: 23499770-23499771 on Build GRCh38
rs1000192661 Chr.3: 23499820-23499820 on Build GRCh38
rs373670059 Chr.3: 23499775-23499775 on Build GRCh38
rs750229816 Chr.3: 23499683-23499683 on Build GRCh38
rs1013881921 Chr.3: 23499796-23499796 on Build GRCh38
rs79454101 Chr.3: 23499765-23499765 on Build GRCh38
rs983961983 Chr.3: 23499704-23499704 on Build GRCh38
rs1000379158 Chr.3: 23499851-23499851 on Build GRCh38
rs780539101 Chr.3: 23499753-23499753 on Build GRCh38
rs191720635 Chr.3: 23499781-23499781 on Build GRCh38
rs769425243 Chr.3: 23499764-23499764 on Build GRCh38
rs147600499 Chr.3: 23499714-23499714 on Build GRCh38
rs374961045 Chr.3: 23499793-23499793 on Build GRCh38
rs747708268 Chr.3: 23499756-23499756 on Build GRCh38
rs184274244 Chr.3: 23499803-23499803 on Build GRCh38
rs905523036 Chr.3: 23499838-23499838 on Build GRCh38
rs929920637 Chr.3: 23499686-23499686 on Build GRCh38
rs779913284 Chr.3: 23499725-23499725 on Build GRCh38

View Full Product Details