Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15017203-15017472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB2
Entrez Gene ID
132864
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
CPE-BP2, CPEB-2, hCPEB-2
UniGene
Hs.656937
Gene Chromosome Location
Chr.4: 15002451-15070153 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_017007733.1 XM_017007734.1 NR_125911.1

Gene Symbol
LOC101929095
Entrez Gene ID
101929095
Gene Name
uncharacterized LOC101929095
Gene Aliases
-
UniGene
Hs.656908
Gene Chromosome Location
Chr.4: 15004942-15427914 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_017007733.1 XM_017007734.1 NR_125911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634532 c.799C>T p.H267Y Chr.4: 15017263-15017263 on Build GRCh38
6634531 c.2110C>T p.H704Y Chr.4: 15017263-15017263 on Build GRCh38
3127349 c.742G>T p.D248Y Chr.4: 15017206-15017206 on Build GRCh38
7087318 c.2092G>T p.D698Y Chr.4: 15017245-15017245 on Build GRCh38
7087319 c.781G>T p.D261Y Chr.4: 15017245-15017245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960881937 Chr.4: 15017392-15017392 on Build GRCh38
rs771036419 Chr.4: 15017252-15017252 on Build GRCh38
rs371070487 Chr.4: 15017301-15017301 on Build GRCh38
rs981124032 Chr.4: 15017368-15017368 on Build GRCh38
rs755975476 Chr.4: 15017292-15017293 on Build GRCh38
rs777662929 Chr.4: 15017302-15017302 on Build GRCh38
rs752387142 Chr.4: 15017311-15017311 on Build GRCh38
rs189490499 Chr.4: 15017314-15017314 on Build GRCh38
rs554609560 Chr.4: 15017433-15017433 on Build GRCh38
rs992005827 Chr.4: 15017431-15017431 on Build GRCh38
rs539739770 Chr.4: 15017435-15017435 on Build GRCh38
rs771450518 Chr.4: 15017278-15017278 on Build GRCh38
rs932070875 Chr.4: 15017288-15017291 on Build GRCh38
rs557282003 Chr.4: 15017238-15017238 on Build GRCh38
rs757450989 Chr.4: 15017407-15017407 on Build GRCh38
rs570297550 Chr.4: 15017244-15017244 on Build GRCh38
rs34609943 Chr.4: 15017276-15017277 on Build GRCh38
rs184997606 Chr.4: 15017309-15017309 on Build GRCh38
rs746431193 Chr.4: 15017262-15017262 on Build GRCh38
rs890442358 Chr.4: 15017299-15017300 on Build GRCh38
rs926572276 Chr.4: 15017384-15017384 on Build GRCh38
rs144630360 Chr.4: 15017239-15017239 on Build GRCh38
rs962109921 Chr.4: 15017292-15017292 on Build GRCh38
rs747053768 Chr.4: 15017245-15017245 on Build GRCh38
rs999953012 Chr.4: 15017364-15017364 on Build GRCh38
rs867566944 Chr.4: 15017338-15017338 on Build GRCh38
rs368796123 Chr.4: 15017283-15017283 on Build GRCh38
rs991185106 Chr.4: 15017421-15017421 on Build GRCh38
rs532841475 Chr.4: 15017293-15017293 on Build GRCh38
rs761192990 Chr.4: 15017249-15017249 on Build GRCh38
rs781648039 Chr.4: 15017257-15017257 on Build GRCh38
rs760243620 Chr.4: 15017299-15017299 on Build GRCh38
rs147297874 Chr.4: 15017303-15017303 on Build GRCh38
rs770190859 Chr.4: 15017264-15017264 on Build GRCh38

View Full Product Details