Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41982279-41982508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF4L1
Entrez Gene ID
285429
Gene Name
DDB1 and CUL4 associated factor 4 like 1
Gene Aliases
WDR21B
UniGene
Hs.213307
Gene Chromosome Location
Chr.4: 41981696-41986467 on Build GRCh38
Transcripts
NM_001029955.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239523 c.498T>A p.S166R Chr.4: 41982290-41982290 on Build GRCh38
6469563 c.569G>T p.W190L Chr.4: 41982361-41982361 on Build GRCh38
1055435 c.487C>T p.R163W Chr.4: 41982279-41982279 on Build GRCh38
268982 c.566C>T p.A189V Chr.4: 41982358-41982358 on Build GRCh38
3603908 c.606A>G p.A202A Chr.4: 41982398-41982398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767569298 Chr.4: 41982356-41982356 on Build GRCh38
rs369697482 Chr.4: 41982433-41982433 on Build GRCh38
rs999352824 Chr.4: 41982371-41982371 on Build GRCh38
rs774728224 Chr.4: 41982458-41982458 on Build GRCh38
rs770165625 Chr.4: 41982316-41982316 on Build GRCh38
rs751721707 Chr.4: 41982344-41982344 on Build GRCh38
rs539660681 Chr.4: 41982461-41982461 on Build GRCh38
rs769274273 Chr.4: 41982434-41982434 on Build GRCh38
rs764486315 Chr.4: 41982334-41982334 on Build GRCh38
rs766690889 Chr.4: 41982488-41982488 on Build GRCh38
rs752513716 Chr.4: 41982375-41982375 on Build GRCh38
rs761023291 Chr.4: 41982486-41982486 on Build GRCh38
rs761795480 Chr.4: 41982352-41982352 on Build GRCh38
rs151240850 Chr.4: 41982359-41982359 on Build GRCh38
rs139209766 Chr.4: 41982378-41982378 on Build GRCh38
rs762325847 Chr.4: 41982474-41982474 on Build GRCh38
rs746896504 Chr.4: 41982401-41982401 on Build GRCh38
rs770360028 Chr.4: 41982412-41982412 on Build GRCh38
rs745550571 Chr.4: 41982424-41982424 on Build GRCh38
rs202183970 Chr.4: 41982426-41982426 on Build GRCh38
rs759128943 Chr.4: 41982307-41982307 on Build GRCh38
rs956323615 Chr.4: 41982411-41982411 on Build GRCh38
rs376010831 Chr.4: 41982420-41982420 on Build GRCh38
rs1051502160 Chr.4: 41982443-41982443 on Build GRCh38
rs769406636 Chr.4: 41982428-41982428 on Build GRCh38
rs750689709 Chr.4: 41982366-41982366 on Build GRCh38
rs777067503 Chr.4: 41982327-41982327 on Build GRCh38
rs372413047 Chr.4: 41982370-41982370 on Build GRCh38
rs372556999 Chr.4: 41982466-41982466 on Build GRCh38
rs775882488 Chr.4: 41982429-41982429 on Build GRCh38
rs777564698 Chr.4: 41982398-41982398 on Build GRCh38
rs775790730 Chr.4: 41982323-41982323 on Build GRCh38
rs932921291 Chr.4: 41982380-41982380 on Build GRCh38
rs900070414 Chr.4: 41982379-41982379 on Build GRCh38
rs767732169 Chr.4: 41982479-41982479 on Build GRCh38
rs756360554 Chr.4: 41982368-41982368 on Build GRCh38
rs773389827 Chr.4: 41982485-41982485 on Build GRCh38

View Full Product Details