Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47976105-47976332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF182
Entrez Gene ID
7569
Gene Name
zinc finger protein 182
Gene Aliases
HHZ150, KOX14, ZNF21, Zfp182
UniGene
Hs.189690
Gene Chromosome Location
Chr.X: 47974851-48003995 on Build GRCh38
Transcripts
NM_001007088.1 NM_001178099.1 NM_006962.1

SNP Detail

SNP ID Chromosome Location
rs781925118 Chr.X: 47976142-47976142 on Build GRCh38
rs782542603 Chr.X: 47976248-47976248 on Build GRCh38
rs368386991 Chr.X: 47976146-47976146 on Build GRCh38
rs782408126 Chr.X: 47976151-47976151 on Build GRCh38
rs781975928 Chr.X: 47976192-47976192 on Build GRCh38
rs984086931 Chr.X: 47976255-47976255 on Build GRCh38
rs782726613 Chr.X: 47976197-47976197 on Build GRCh38
rs952532293 Chr.X: 47976179-47976179 on Build GRCh38
rs975232154 Chr.X: 47976139-47976139 on Build GRCh38
rs782064823 Chr.X: 47976234-47976234 on Build GRCh38
rs782094423 Chr.X: 47976193-47976193 on Build GRCh38
rs781802034 Chr.X: 47976226-47976226 on Build GRCh38
rs781883797 Chr.X: 47976268-47976268 on Build GRCh38

View Full Product Details