Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 129442766-129443027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARX2
Entrez Gene ID
8538
Gene Name
BARX homeobox 2
Gene Aliases
-
UniGene
Hs.591944
Gene Chromosome Location
Chr.11: 129375237-129452279 on Build GRCh38
Transcripts
NM_003658.4 XM_011543043.1 XM_011543044.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357291 c.499G>T p.A167S Chr.11: 129442845-129442845 on Build GRCh38
1127849 c.566A>G p.K189R Chr.11: 129442912-129442912 on Build GRCh38
6280292 c.550C>T p.R184C Chr.11: 129442896-129442896 on Build GRCh38
231829 c.529G>A p.V177M Chr.11: 129442875-129442875 on Build GRCh38
139935 c.493G>A p.D165N Chr.11: 129442839-129442839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775040046 Chr.11: 129442808-129442808 on Build GRCh38
rs1046602249 Chr.11: 129442875-129442875 on Build GRCh38
rs906881234 Chr.11: 129442904-129442904 on Build GRCh38
rs373737561 Chr.11: 129442815-129442815 on Build GRCh38
rs983358283 Chr.11: 129442822-129442822 on Build GRCh38
rs367573574 Chr.11: 129442844-129442844 on Build GRCh38
rs772635685 Chr.11: 129442964-129442964 on Build GRCh38
rs542177685 Chr.11: 129442789-129442789 on Build GRCh38
rs756228193 Chr.11: 129442867-129442867 on Build GRCh38
rs761331239 Chr.11: 129442830-129442830 on Build GRCh38
rs776021147 Chr.11: 129442828-129442828 on Build GRCh38
rs748965066 Chr.11: 129442913-129442913 on Build GRCh38
rs747366539 Chr.11: 129442803-129442804 on Build GRCh38
rs545281008 Chr.11: 129442996-129442996 on Build GRCh38
rs771352228 Chr.11: 129442803-129442803 on Build GRCh38
rs777662666 Chr.11: 129442877-129442877 on Build GRCh38
rs761082396 Chr.11: 129442990-129442990 on Build GRCh38
rs866130127 Chr.11: 129442987-129442987 on Build GRCh38
rs746220345 Chr.11: 129442955-129442955 on Build GRCh38
rs559226837 Chr.11: 129442988-129442988 on Build GRCh38
rs942100678 Chr.11: 129442811-129442812 on Build GRCh38
rs750219485 Chr.11: 129442839-129442839 on Build GRCh38
rs765120324 Chr.11: 129442835-129442835 on Build GRCh38
rs776360676 Chr.11: 129442819-129442819 on Build GRCh38
rs757392538 Chr.11: 129442907-129442907 on Build GRCh38
rs370794313 Chr.11: 129442862-129442862 on Build GRCh38
rs147038314 Chr.11: 129442888-129442888 on Build GRCh38
rs528418656 Chr.11: 129442993-129442993 on Build GRCh38
rs766411728 Chr.11: 129442854-129442854 on Build GRCh38
rs774544076 Chr.11: 129442804-129442804 on Build GRCh38
rs370281521 Chr.11: 129442807-129442807 on Build GRCh38
rs373893156 Chr.11: 129442936-129442936 on Build GRCh38
rs138172946 Chr.11: 129442897-129442897 on Build GRCh38
rs760175465 Chr.11: 129442811-129442811 on Build GRCh38
rs749408863 Chr.11: 129442790-129442790 on Build GRCh38
rs760079163 Chr.11: 129442805-129442805 on Build GRCh38
rs897957750 Chr.11: 129442832-129442832 on Build GRCh38

View Full Product Details