Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5508191-5508448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBQLN3
Entrez Gene ID
50613
Gene Name
ubiquilin 3
Gene Aliases
TUP-1
UniGene
Hs.189184
Gene Chromosome Location
Chr.11: 5507300-5509993 on Build GRCh38
Transcripts
NM_017481.2 XM_011520144.2 XM_011520145.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
324159 c.1218G>T p.R406S Chr.11: 5508341-5508341 on Build GRCh38
1475511 c.1207A>C p.I403L Chr.11: 5508352-5508352 on Build GRCh38
5372695 c.1325C>T p.P442L Chr.11: 5508234-5508234 on Build GRCh38
5747214 c.1250C>T p.T417I Chr.11: 5508309-5508309 on Build GRCh38
1492589 c.1272T>C p.G424G Chr.11: 5508287-5508287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751135826 Chr.11: 5508420-5508420 on Build GRCh38
rs769749020 Chr.11: 5508367-5508367 on Build GRCh38
rs866029134 Chr.11: 5508382-5508382 on Build GRCh38
rs759817450 Chr.11: 5508311-5508311 on Build GRCh38
rs754356380 Chr.11: 5508412-5508412 on Build GRCh38
rs115774750 Chr.11: 5508289-5508289 on Build GRCh38
rs559044353 Chr.11: 5508215-5508215 on Build GRCh38
rs768558010 Chr.11: 5508241-5508241 on Build GRCh38
rs762294011 Chr.11: 5508417-5508417 on Build GRCh38
rs753077189 Chr.11: 5508313-5508313 on Build GRCh38
rs766617498 Chr.11: 5508301-5508302 on Build GRCh38
rs756500901 Chr.11: 5508221-5508221 on Build GRCh38
rs749238008 Chr.11: 5508376-5508376 on Build GRCh38
rs757904611 Chr.11: 5508329-5508329 on Build GRCh38
rs567893872 Chr.11: 5508290-5508290 on Build GRCh38
rs369978259 Chr.11: 5508232-5508232 on Build GRCh38
rs868733929 Chr.11: 5508369-5508369 on Build GRCh38
rs779488363 Chr.11: 5508331-5508331 on Build GRCh38
rs774615319 Chr.11: 5508294-5508294 on Build GRCh38
rs996260519 Chr.11: 5508411-5508411 on Build GRCh38
rs756621908 Chr.11: 5508216-5508216 on Build GRCh38
rs771035029 Chr.11: 5508377-5508377 on Build GRCh38
rs777741072 Chr.11: 5508368-5508368 on Build GRCh38
rs776572297 Chr.11: 5508256-5508256 on Build GRCh38
rs754690474 Chr.11: 5508342-5508342 on Build GRCh38
rs751767264 Chr.11: 5508213-5508213 on Build GRCh38
rs999501348 Chr.11: 5508330-5508330 on Build GRCh38
rs2234452 Chr.11: 5508410-5508410 on Build GRCh38
rs773124058 Chr.11: 5508287-5508287 on Build GRCh38
rs760819086 Chr.11: 5508213-5508213 on Build GRCh38
rs748040510 Chr.11: 5508264-5508264 on Build GRCh38
rs145187997 Chr.11: 5508400-5508400 on Build GRCh38
rs759590961 Chr.11: 5508358-5508381 on Build GRCh38
rs769873541 Chr.11: 5508271-5508271 on Build GRCh38
rs774303947 Chr.11: 5508381-5508381 on Build GRCh38
rs998972912 Chr.11: 5508406-5508406 on Build GRCh38
rs2234453 Chr.11: 5508211-5508211 on Build GRCh38
rs767807168 Chr.11: 5508312-5508312 on Build GRCh38
rs890971555 Chr.11: 5508286-5508286 on Build GRCh38
rs200573647 Chr.11: 5508365-5508365 on Build GRCh38
rs143750640 Chr.11: 5508222-5508222 on Build GRCh38
rs754344419 Chr.11: 5508327-5508327 on Build GRCh38
rs899803218 Chr.11: 5508335-5508335 on Build GRCh38
rs202058718 Chr.11: 5508399-5508399 on Build GRCh38
rs746669696 Chr.11: 5508336-5508336 on Build GRCh38
rs756566967 Chr.11: 5508318-5508318 on Build GRCh38
rs373862522 Chr.11: 5508419-5508419 on Build GRCh38
rs764619503 Chr.11: 5508321-5508321 on Build GRCh38
rs759700974 Chr.11: 5508386-5508386 on Build GRCh38
rs776644554 Chr.11: 5508340-5508341 on Build GRCh38

View Full Product Details