Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143323928-143324201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOP1MT
Entrez Gene ID
116447
Gene Name
topoisomerase (DNA) I, mitochondrial
Gene Aliases
-
UniGene
Hs.528574
Gene Chromosome Location
Chr.8: 143309324-143359977 on Build GRCh38
Transcripts
NM_001258446.1 NM_001258447.1 NM_052963.2 XM_005250781.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781395 c.898G>A p.D300N Chr.8: 143324061-143324061 on Build GRCh38
3778961 c.931C>T p.R311W Chr.8: 143324028-143324028 on Build GRCh38
4562691 c.939G>A p.V313V Chr.8: 143324020-143324020 on Build GRCh38
1096928 c.860G>A p.R287Q Chr.8: 143324099-143324099 on Build GRCh38
749790 c.952A>G p.I318V Chr.8: 143324007-143324007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368070146 Chr.8: 143323984-143323984 on Build GRCh38
rs148151254 Chr.8: 143324033-143324033 on Build GRCh38
rs772059174 Chr.8: 143324011-143324011 on Build GRCh38
rs771755999 Chr.8: 143324078-143324078 on Build GRCh38
rs759235799 Chr.8: 143324023-143324023 on Build GRCh38
rs143769145 Chr.8: 143324004-143324004 on Build GRCh38
rs940604315 Chr.8: 143324120-143324120 on Build GRCh38
rs776656218 Chr.8: 143324167-143324167 on Build GRCh38
rs769133631 Chr.8: 143324144-143324144 on Build GRCh38
rs375561204 Chr.8: 143324028-143324028 on Build GRCh38
rs749453847 Chr.8: 143324156-143324156 on Build GRCh38
rs764945190 Chr.8: 143324024-143324024 on Build GRCh38
rs770602484 Chr.8: 143324085-143324085 on Build GRCh38
rs761466166 Chr.8: 143324010-143324010 on Build GRCh38
rs201977682 Chr.8: 143324048-143324048 on Build GRCh38
rs969037830 Chr.8: 143324069-143324069 on Build GRCh38
rs373121260 Chr.8: 143324063-143324063 on Build GRCh38
rs112528843 Chr.8: 143323992-143323992 on Build GRCh38
rs370926126 Chr.8: 143324162-143324162 on Build GRCh38
rs964007864 Chr.8: 143324071-143324071 on Build GRCh38
rs144092447 Chr.8: 143324079-143324079 on Build GRCh38
rs146854602 Chr.8: 143324142-143324142 on Build GRCh38
rs781461370 Chr.8: 143323998-143323998 on Build GRCh38
rs759071083 Chr.8: 143324096-143324096 on Build GRCh38
rs1031626496 Chr.8: 143324166-143324166 on Build GRCh38
rs376848972 Chr.8: 143324149-143324149 on Build GRCh38
rs374704852 Chr.8: 143323974-143323974 on Build GRCh38
rs1000613727 Chr.8: 143324158-143324158 on Build GRCh38
rs147333453 Chr.8: 143324106-143324106 on Build GRCh38
rs781227767 Chr.8: 143324056-143324056 on Build GRCh38
rs577773438 Chr.8: 143324066-143324066 on Build GRCh38
rs756119309 Chr.8: 143324148-143324148 on Build GRCh38
rs368640703 Chr.8: 143323948-143323948 on Build GRCh38
rs766056085 Chr.8: 143323957-143323957 on Build GRCh38
rs772952475 Chr.8: 143324014-143324014 on Build GRCh38
rs987068727 Chr.8: 143324054-143324054 on Build GRCh38
rs763441442 Chr.8: 143324100-143324100 on Build GRCh38
rs774942791 Chr.8: 143324099-143324099 on Build GRCh38
rs753138890 Chr.8: 143324027-143324027 on Build GRCh38
rs886382708 Chr.8: 143323972-143323972 on Build GRCh38
rs760294605 Chr.8: 143324017-143324017 on Build GRCh38
rs758260106 Chr.8: 143324157-143324157 on Build GRCh38
rs919986376 Chr.8: 143324082-143324082 on Build GRCh38
rs907695758 Chr.8: 143324119-143324119 on Build GRCh38
rs745429425 Chr.8: 143323981-143323982 on Build GRCh38
rs374591550 Chr.8: 143324147-143324147 on Build GRCh38
rs776468325 Chr.8: 143324022-143324022 on Build GRCh38
rs755449565 Chr.8: 143323973-143323973 on Build GRCh38
rs753300396 Chr.8: 143323983-143323983 on Build GRCh38
rs747979751 Chr.8: 143324074-143324074 on Build GRCh38
rs752005241 Chr.8: 143324037-143324037 on Build GRCh38
rs988812844 Chr.8: 143324141-143324141 on Build GRCh38
rs765789984 Chr.8: 143324019-143324019 on Build GRCh38
rs200872473 Chr.8: 143323993-143323993 on Build GRCh38
rs369652527 Chr.8: 143324036-143324036 on Build GRCh38
rs752158768 Chr.8: 143324109-143324109 on Build GRCh38
rs756349997 Chr.8: 143324064-143324064 on Build GRCh38
rs117859508 Chr.8: 143323963-143323963 on Build GRCh38
rs912571568 Chr.8: 143324067-143324067 on Build GRCh38
rs371532680 Chr.8: 143323988-143323988 on Build GRCh38
rs774161309 Chr.8: 143324005-143324005 on Build GRCh38
rs776314009 Chr.8: 143324086-143324086 on Build GRCh38
rs760045915 Chr.8: 143323949-143323949 on Build GRCh38

View Full Product Details