Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84191582-84191839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD2
Entrez Gene ID
161931
Gene Name
adenosine deaminase domain containing 2
Gene Aliases
TENRL
UniGene
Hs.8977
Gene Chromosome Location
Chr.16: 84191117-84197166 on Build GRCh38
Transcripts
NM_001145400.1 NM_139174.3 XM_005255814.1 XM_011522919.1 XM_011522920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645926 c.362C>T p.T121M Chr.16: 84191592-84191592 on Build GRCh38
5958345 c.360C>G p.L120L Chr.16: 84191590-84191590 on Build GRCh38
5657729 c.400C>T p.R134W Chr.16: 84191630-84191630 on Build GRCh38
4565164 c.418_418+1GG>AA p.? Chr.16: 84191648-84191649 on Build GRCh38
5498231 c.371C>T p.A124V Chr.16: 84191601-84191601 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913209333 Chr.16: 84191642-84191642 on Build GRCh38
rs763801209 Chr.16: 84191648-84191648 on Build GRCh38
rs531532957 Chr.16: 84191796-84191796 on Build GRCh38
rs759351236 Chr.16: 84191616-84191616 on Build GRCh38
rs964266240 Chr.16: 84191804-84191804 on Build GRCh38
rs760082026 Chr.16: 84191661-84191661 on Build GRCh38
rs547879944 Chr.16: 84191641-84191641 on Build GRCh38
rs747180214 Chr.16: 84191727-84191727 on Build GRCh38
rs899444918 Chr.16: 84191743-84191743 on Build GRCh38
rs775029443 Chr.16: 84191626-84191626 on Build GRCh38
rs979205609 Chr.16: 84191787-84191787 on Build GRCh38
rs776651747 Chr.16: 84191614-84191614 on Build GRCh38
rs896001401 Chr.16: 84191768-84191768 on Build GRCh38
rs942542235 Chr.16: 84191676-84191676 on Build GRCh38
rs569964097 Chr.16: 84191699-84191699 on Build GRCh38
rs1039758862 Chr.16: 84191755-84191755 on Build GRCh38
rs1014337653 Chr.16: 84191769-84191769 on Build GRCh38
rs556455353 Chr.16: 84191609-84191609 on Build GRCh38
rs778155277 Chr.16: 84191725-84191725 on Build GRCh38
rs1037951406 Chr.16: 84191742-84191742 on Build GRCh38
rs545701645 Chr.16: 84191665-84191665 on Build GRCh38
rs922441850 Chr.16: 84191713-84191713 on Build GRCh38
rs1001799514 Chr.16: 84191754-84191754 on Build GRCh38
rs527684587 Chr.16: 84191718-84191718 on Build GRCh38
rs754203074 Chr.16: 84191678-84191678 on Build GRCh38
rs780888870 Chr.16: 84191752-84191752 on Build GRCh38
rs796919350 Chr.16: 84191721-84191722 on Build GRCh38
rs766707473 Chr.16: 84191639-84191639 on Build GRCh38
rs973576788 Chr.16: 84191657-84191657 on Build GRCh38
rs561442926 Chr.16: 84191722-84191722 on Build GRCh38
rs1020406419 Chr.16: 84191783-84191783 on Build GRCh38
rs757793881 Chr.16: 84191820-84191820 on Build GRCh38
rs111685508 Chr.16: 84191766-84191766 on Build GRCh38
rs1051711705 Chr.16: 84191772-84191772 on Build GRCh38
rs143947465 Chr.16: 84191706-84191706 on Build GRCh38
rs967402534 Chr.16: 84191784-84191784 on Build GRCh38
rs200040548 Chr.16: 84191624-84191624 on Build GRCh38
rs766616095 Chr.16: 84191662-84191662 on Build GRCh38
rs138132588 Chr.16: 84191723-84191730 on Build GRCh38
rs8049081 Chr.16: 84191721-84191721 on Build GRCh38
rs993123283 Chr.16: 84191712-84191712 on Build GRCh38
rs1034424850 Chr.16: 84191602-84191602 on Build GRCh38
rs190742693 Chr.16: 84191732-84191732 on Build GRCh38
rs922410599 Chr.16: 84191813-84191813 on Build GRCh38
rs377713078 Chr.16: 84191658-84191658 on Build GRCh38
rs1004567031 Chr.16: 84191812-84191812 on Build GRCh38
rs367685601 Chr.16: 84191765-84191765 on Build GRCh38
rs1014702043 Chr.16: 84191601-84191601 on Build GRCh38
rs751142260 Chr.16: 84191656-84191656 on Build GRCh38
rs975373777 Chr.16: 84191810-84191810 on Build GRCh38

View Full Product Details