Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44354593-44354850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM2
Entrez Gene ID
7226
Gene Name
transient receptor potential cation channel subfamily M member 2
Gene Aliases
EREG1, KNP3, LTRPC2, LTrpC-2, NUDT9H, NUDT9L1, TRPC7
UniGene
Hs.369759
Gene Chromosome Location
Chr.21: 44350112-44443081 on Build GRCh38
Transcripts
NM_001320350.1 NM_001320351.1 NM_003307.3 NR_038257.1 XM_005261171.3 XM_011529736.2 XM_017028456.1 XM_017028457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6256483 c.179G>T p.S60I Chr.21: 44354661-44354661 on Build GRCh38
7425277 c.218T>A p.V73E Chr.21: 44354700-44354700 on Build GRCh38
7249777 c.224T>C p.F75S Chr.21: 44354706-44354706 on Build GRCh38
3551356 c.190C>T p.P64S Chr.21: 44354672-44354672 on Build GRCh38
3972802 c.183G>A p.S61S Chr.21: 44354665-44354665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965520627 Chr.21: 44354811-44354811 on Build GRCh38
rs780421280 Chr.21: 44354663-44354663 on Build GRCh38
rs374742193 Chr.21: 44354781-44354781 on Build GRCh38
rs746035733 Chr.21: 44354647-44354647 on Build GRCh38
rs764007734 Chr.21: 44354745-44354745 on Build GRCh38
rs748355932 Chr.21: 44354687-44354690 on Build GRCh38
rs1052396788 Chr.21: 44354684-44354684 on Build GRCh38
rs759178437 Chr.21: 44354699-44354699 on Build GRCh38
rs45568434 Chr.21: 44354794-44354794 on Build GRCh38
rs9979269 Chr.21: 44354783-44354783 on Build GRCh38
rs751617397 Chr.21: 44354614-44354614 on Build GRCh38
rs756322450 Chr.21: 44354785-44354785 on Build GRCh38
rs371708768 Chr.21: 44354660-44354660 on Build GRCh38
rs770494214 Chr.21: 44354693-44354693 on Build GRCh38
rs1015182595 Chr.21: 44354803-44354803 on Build GRCh38
rs780326525 Chr.21: 44354786-44354786 on Build GRCh38
rs892553080 Chr.21: 44354685-44354685 on Build GRCh38
rs199640840 Chr.21: 44354746-44354746 on Build GRCh38
rs772912846 Chr.21: 44354672-44354672 on Build GRCh38
rs547075317 Chr.21: 44354677-44354677 on Build GRCh38
rs200293453 Chr.21: 44354664-44354664 on Build GRCh38
rs769258683 Chr.21: 44354665-44354665 on Build GRCh38
rs367841933 Chr.21: 44354645-44354645 on Build GRCh38
rs141640522 Chr.21: 44354732-44354732 on Build GRCh38
rs761886405 Chr.21: 44354774-44354774 on Build GRCh38
rs1027229961 Chr.21: 44354782-44354782 on Build GRCh38
rs764971860 Chr.21: 44354706-44354706 on Build GRCh38
rs776157534 Chr.21: 44354698-44354698 on Build GRCh38
rs369333401 Chr.21: 44354790-44354790 on Build GRCh38

View Full Product Details