Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56121089-56121338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H10
Entrez Gene ID
84872
Gene Name
zinc finger CCCH-type containing 10
Gene Aliases
ZC3HDC10
UniGene
Hs.632706
Gene Chromosome Location
Chr.12: 56118220-56122497 on Build GRCh38
Transcripts
NM_001303124.1 NM_001303125.1 NM_032786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755850 c.628C>T p.R210W Chr.12: 56121190-56121190 on Build GRCh38
4043480 c.670T>C p.S224P Chr.12: 56121232-56121232 on Build GRCh38
1270740 c.569_571delATG p.D191delD Chr.12: 56121131-56121133 on Build GRCh38
4972149 c.553C>T p.R185* Chr.12: 56121115-56121115 on Build GRCh38
248418 c.629G>A p.R210Q Chr.12: 56121191-56121191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367724913 Chr.12: 56121233-56121233 on Build GRCh38
rs1003184641 Chr.12: 56121113-56121113 on Build GRCh38
rs760848989 Chr.12: 56121116-56121116 on Build GRCh38
rs745537120 Chr.12: 56121108-56121108 on Build GRCh38
rs907606088 Chr.12: 56121312-56121312 on Build GRCh38
rs548618961 Chr.12: 56121212-56121212 on Build GRCh38
rs867346846 Chr.12: 56121208-56121208 on Build GRCh38
rs748473927 Chr.12: 56121260-56121260 on Build GRCh38
rs145833211 Chr.12: 56121118-56121118 on Build GRCh38
rs755399288 Chr.12: 56121309-56121309 on Build GRCh38
rs777409875 Chr.12: 56121137-56121137 on Build GRCh38
rs753750097 Chr.12: 56121119-56121119 on Build GRCh38
rs754766642 Chr.12: 56121127-56121127 on Build GRCh38
rs749799266 Chr.12: 56121180-56121180 on Build GRCh38
rs763691100 Chr.12: 56121231-56121231 on Build GRCh38
rs755505028 Chr.12: 56121257-56121257 on Build GRCh38
rs773590176 Chr.12: 56121207-56121207 on Build GRCh38
rs772253304 Chr.12: 56121305-56121305 on Build GRCh38
rs774858587 Chr.12: 56121129-56121129 on Build GRCh38
rs762752571 Chr.12: 56121221-56121221 on Build GRCh38
rs147799715 Chr.12: 56121153-56121153 on Build GRCh38
rs200128733 Chr.12: 56121167-56121167 on Build GRCh38
rs1000598312 Chr.12: 56121174-56121174 on Build GRCh38
rs74094539 Chr.12: 56121156-56121156 on Build GRCh38
rs371573767 Chr.12: 56121181-56121181 on Build GRCh38
rs531554160 Chr.12: 56121190-56121190 on Build GRCh38
rs756743578 Chr.12: 56121143-56121143 on Build GRCh38
rs758111329 Chr.12: 56121131-56121131 on Build GRCh38
rs955576519 Chr.12: 56121243-56121243 on Build GRCh38
rs996103555 Chr.12: 56121240-56121240 on Build GRCh38
rs371248458 Chr.12: 56121258-56121258 on Build GRCh38
rs780616920 Chr.12: 56121245-56121245 on Build GRCh38
rs754358164 Chr.12: 56121255-56121255 on Build GRCh38
rs752425072 Chr.12: 56121219-56121219 on Build GRCh38
rs772595730 Chr.12: 56121263-56121263 on Build GRCh38
rs141980139 Chr.12: 56121109-56121109 on Build GRCh38
rs769043530 Chr.12: 56121182-56121182 on Build GRCh38
rs756760763 Chr.12: 56121236-56121236 on Build GRCh38
rs752330809 Chr.12: 56121130-56121130 on Build GRCh38
rs760797657 Chr.12: 56121209-56121209 on Build GRCh38
rs894927183 Chr.12: 56121154-56121154 on Build GRCh38
rs374457746 Chr.12: 56121211-56121211 on Build GRCh38
rs754965926 Chr.12: 56121113-56121115 on Build GRCh38
rs772361984 Chr.12: 56121201-56121201 on Build GRCh38
rs983329036 Chr.12: 56121280-56121280 on Build GRCh38

View Full Product Details