Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100430411-100430638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEPCE
Entrez Gene ID
56257
Gene Name
methylphosphate capping enzyme
Gene Aliases
BCDIN3
UniGene
Hs.178011
Gene Chromosome Location
Chr.7: 100428790-100434126 on Build GRCh38
Transcripts
NM_001194990.1 NM_001194991.1 NM_001194992.1 NM_019606.5 XM_011516410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
245659 c.457T>A p.C153S Chr.7: 100430475-100430475 on Build GRCh38
5588277 c.601C>G p.L201V Chr.7: 100430619-100430619 on Build GRCh38
4701756 c.427delG p.G145fs*11 Chr.7: 100430445-100430445 on Build GRCh38
5649560 c.409C>G p.Q137E Chr.7: 100430427-100430427 on Build GRCh38
3080172 c.418C>T p.R140W Chr.7: 100430436-100430436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056116897 Chr.7: 100430618-100430618 on Build GRCh38
rs779120161 Chr.7: 100430457-100430457 on Build GRCh38
rs762883519 Chr.7: 100430522-100430522 on Build GRCh38
rs764787290 Chr.7: 100430517-100430517 on Build GRCh38
rs779891739 Chr.7: 100430435-100430435 on Build GRCh38
rs760899371 Chr.7: 100430480-100430480 on Build GRCh38
rs771795632 Chr.7: 100430474-100430474 on Build GRCh38
rs749930549 Chr.7: 100430521-100430521 on Build GRCh38
rs751396535 Chr.7: 100430546-100430546 on Build GRCh38
rs1003653921 Chr.7: 100430444-100430444 on Build GRCh38
rs979057742 Chr.7: 100430555-100430555 on Build GRCh38
rs144659071 Chr.7: 100430576-100430576 on Build GRCh38
rs760012501 Chr.7: 100430445-100430445 on Build GRCh38
rs761520441 Chr.7: 100430591-100430591 on Build GRCh38
rs373454923 Chr.7: 100430483-100430483 on Build GRCh38
rs886593717 Chr.7: 100430430-100430430 on Build GRCh38
rs751247589 Chr.7: 100430442-100430442 on Build GRCh38
rs761506452 Chr.7: 100430502-100430502 on Build GRCh38
rs758320148 Chr.7: 100430432-100430432 on Build GRCh38
rs766250970 Chr.7: 100430536-100430536 on Build GRCh38
rs375858105 Chr.7: 100430553-100430553 on Build GRCh38
rs1007677513 Chr.7: 100430449-100430449 on Build GRCh38
rs776184751 Chr.7: 100430494-100430494 on Build GRCh38
rs1034398184 Chr.7: 100430498-100430498 on Build GRCh38
rs769371814 Chr.7: 100430619-100430619 on Build GRCh38
rs754691853 Chr.7: 100430551-100430551 on Build GRCh38
rs142061761 Chr.7: 100430574-100430574 on Build GRCh38
rs777041063 Chr.7: 100430587-100430587 on Build GRCh38
rs779134499 Chr.7: 100430566-100430566 on Build GRCh38
rs745881376 Chr.7: 100430569-100430569 on Build GRCh38
rs756561147 Chr.7: 100430446-100430446 on Build GRCh38
rs754062405 Chr.7: 100430554-100430554 on Build GRCh38
rs769103963 Chr.7: 100430582-100430582 on Build GRCh38
rs776864380 Chr.7: 100430444-100430445 on Build GRCh38
rs6975729 Chr.7: 100430564-100430564 on Build GRCh38
rs1019105876 Chr.7: 100430455-100430455 on Build GRCh38
rs768949839 Chr.7: 100430585-100430585 on Build GRCh38
rs770129976 Chr.7: 100430578-100430579 on Build GRCh38
rs776008948 Chr.7: 100430478-100430478 on Build GRCh38
rs777812010 Chr.7: 100430447-100430447 on Build GRCh38
rs746440132 Chr.7: 100430464-100430464 on Build GRCh38

View Full Product Details