Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110672994-110673263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA3
Entrez Gene ID
3738
Gene Name
potassium voltage-gated channel subfamily A member 3
Gene Aliases
HGK5, HLK3, HPCN3, HUKIII, KV1.3, MK3, PCN3
UniGene
Hs.169948
Gene Chromosome Location
Chr.1: 110653560-110675033 on Build GRCh38
Transcripts
NM_002232.4 NR_109845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843036 c.1606G>A p.G536S Chr.1: 110673204-110673204 on Build GRCh38
6690441 c.1550A>G p.K517R Chr.1: 110673260-110673260 on Build GRCh38
3718384 c.1626C>T p.F542F Chr.1: 110673184-110673184 on Build GRCh38
347923 c.1572G>C p.L524L Chr.1: 110673238-110673238 on Build GRCh38
893702 c.1621G>A p.A541T Chr.1: 110673189-110673189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773212851 Chr.1: 110673107-110673107 on Build GRCh38
rs138887050 Chr.1: 110673232-110673232 on Build GRCh38
rs902667859 Chr.1: 110673181-110673181 on Build GRCh38
rs779056951 Chr.1: 110673219-110673219 on Build GRCh38
rs746090912 Chr.1: 110673222-110673222 on Build GRCh38
rs771014106 Chr.1: 110673155-110673155 on Build GRCh38
rs946802393 Chr.1: 110673150-110673150 on Build GRCh38
rs752649641 Chr.1: 110673031-110673031 on Build GRCh38
rs772195154 Chr.1: 110673106-110673106 on Build GRCh38
rs747145696 Chr.1: 110673207-110673207 on Build GRCh38
rs868826912 Chr.1: 110673092-110673092 on Build GRCh38
rs770835476 Chr.1: 110673100-110673100 on Build GRCh38
rs756586190 Chr.1: 110673190-110673190 on Build GRCh38
rs201084914 Chr.1: 110673064-110673064 on Build GRCh38
rs748661099 Chr.1: 110673205-110673205 on Build GRCh38
rs775628549 Chr.1: 110673194-110673194 on Build GRCh38
rs199848826 Chr.1: 110673203-110673203 on Build GRCh38
rs376424861 Chr.1: 110673030-110673030 on Build GRCh38
rs751268655 Chr.1: 110673047-110673047 on Build GRCh38
rs376883477 Chr.1: 110673079-110673079 on Build GRCh38
rs756902323 Chr.1: 110673050-110673050 on Build GRCh38
rs749566129 Chr.1: 110673071-110673071 on Build GRCh38
rs145738982 Chr.1: 110673174-110673174 on Build GRCh38
rs568444948 Chr.1: 110673204-110673204 on Build GRCh38
rs763013710 Chr.1: 110673239-110673239 on Build GRCh38
rs749033932 Chr.1: 110673077-110673077 on Build GRCh38
rs990829035 Chr.1: 110673110-110673110 on Build GRCh38
rs1027762969 Chr.1: 110673160-110673160 on Build GRCh38
rs776785948 Chr.1: 110673218-110673218 on Build GRCh38
rs751072580 Chr.1: 110673200-110673200 on Build GRCh38
rs375722863 Chr.1: 110673039-110673039 on Build GRCh38
rs915300937 Chr.1: 110673116-110673116 on Build GRCh38
rs373986871 Chr.1: 110673073-110673073 on Build GRCh38
rs753131399 Chr.1: 110673158-110673158 on Build GRCh38
rs1040786526 Chr.1: 110673062-110673062 on Build GRCh38
rs762924096 Chr.1: 110673198-110673198 on Build GRCh38
rs763699947 Chr.1: 110673199-110673199 on Build GRCh38
rs758865322 Chr.1: 110673206-110673206 on Build GRCh38
rs994008309 Chr.1: 110673097-110673097 on Build GRCh38
rs747266463 Chr.1: 110673128-110673128 on Build GRCh38
rs777410901 Chr.1: 110673040-110673040 on Build GRCh38
rs748646757 Chr.1: 110673088-110673088 on Build GRCh38
rs769188853 Chr.1: 110673127-110673129 on Build GRCh38
rs775356636 Chr.1: 110673233-110673233 on Build GRCh38
rs141089852 Chr.1: 110673184-110673184 on Build GRCh38

View Full Product Details