Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40022235-40022508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED24
Entrez Gene ID
9862
Gene Name
mediator complex subunit 24
Gene Aliases
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
UniGene
Hs.462983
Gene Chromosome Location
Chr.17: 40019097-40054636 on Build GRCh38
Transcripts
NM_001079518.1 NM_001267797.1 NM_014815.3 NR_052017.1 XM_005257874.2 XM_006722204.2 XM_006722206.2 XM_006722207.2 XM_011525529.2 XM_011525530.2 XM_011525531.2 XM_011525532.2 XM_011525533.2 XM_011525534.2 XM_011525535.2 XM_011525536.2 XM_011525538.2 XM_017025458.1 XM_017025459.1 XM_017025460.1 XM_017025461.1 XM_017025462.1 XM_017025463.1 XM_017025464.1 XM_017025465.1 XM_017025466.1 XM_017025467.1 XM_017025468.1 XM_017025469.1 XM_017025470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
978762 c.2512G>A p.E838K Chr.17: 40022405-40022405 on Build GRCh38
288083 c.2478A>C p.G826G Chr.17: 40022439-40022439 on Build GRCh38
978764 c.2511C>T p.R837R Chr.17: 40022406-40022406 on Build GRCh38
4958884 c.2475G>A p.K825K Chr.17: 40022442-40022442 on Build GRCh38
4617757 c.2490C>T p.T830T Chr.17: 40022427-40022427 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045329723 Chr.17: 40022275-40022275 on Build GRCh38
rs376898440 Chr.17: 40022485-40022485 on Build GRCh38
rs765780838 Chr.17: 40022480-40022480 on Build GRCh38
rs149459087 Chr.17: 40022426-40022426 on Build GRCh38
rs761022928 Chr.17: 40022451-40022451 on Build GRCh38
rs183983029 Chr.17: 40022377-40022377 on Build GRCh38
rs767808390 Chr.17: 40022373-40022373 on Build GRCh38
rs536794729 Chr.17: 40022371-40022371 on Build GRCh38
rs754369451 Chr.17: 40022378-40022378 on Build GRCh38
rs552966224 Chr.17: 40022406-40022406 on Build GRCh38
rs974044040 Chr.17: 40022393-40022393 on Build GRCh38
rs904851162 Chr.17: 40022320-40022320 on Build GRCh38
rs891204758 Chr.17: 40022314-40022314 on Build GRCh38
rs751531174 Chr.17: 40022426-40022426 on Build GRCh38
rs143907532 Chr.17: 40022465-40022465 on Build GRCh38
rs767387725 Chr.17: 40022355-40022355 on Build GRCh38
rs370310367 Chr.17: 40022487-40022487 on Build GRCh38
rs758098673 Chr.17: 40022359-40022359 on Build GRCh38
rs185766761 Chr.17: 40022257-40022257 on Build GRCh38
rs949580213 Chr.17: 40022428-40022428 on Build GRCh38
rs371797692 Chr.17: 40022478-40022478 on Build GRCh38
rs919939546 Chr.17: 40022405-40022405 on Build GRCh38
rs764606546 Chr.17: 40022383-40022383 on Build GRCh38
rs918101968 Chr.17: 40022260-40022260 on Build GRCh38
rs759614226 Chr.17: 40022353-40022353 on Build GRCh38
rs1025153796 Chr.17: 40022382-40022383 on Build GRCh38
rs757922584 Chr.17: 40022397-40022397 on Build GRCh38
rs372694934 Chr.17: 40022274-40022274 on Build GRCh38
rs371799904 Chr.17: 40022343-40022343 on Build GRCh38
rs777161039 Chr.17: 40022477-40022477 on Build GRCh38
rs945187588 Chr.17: 40022277-40022277 on Build GRCh38
rs377656209 Chr.17: 40022376-40022376 on Build GRCh38
rs371978898 Chr.17: 40022369-40022369 on Build GRCh38
rs1017560124 Chr.17: 40022381-40022381 on Build GRCh38
rs180904097 Chr.17: 40022370-40022370 on Build GRCh38
rs3213762 Chr.17: 40022374-40022374 on Build GRCh38
rs755594371 Chr.17: 40022382-40022382 on Build GRCh38
rs201314858 Chr.17: 40022408-40022408 on Build GRCh38
rs529431580 Chr.17: 40022352-40022352 on Build GRCh38
rs1051589251 Chr.17: 40022289-40022289 on Build GRCh38
rs551743592 Chr.17: 40022372-40022372 on Build GRCh38
rs748261743 Chr.17: 40022430-40022430 on Build GRCh38
rs547275352 Chr.17: 40022325-40022325 on Build GRCh38
rs774346788 Chr.17: 40022300-40022300 on Build GRCh38
rs970425242 Chr.17: 40022384-40022384 on Build GRCh38
rs753624032 Chr.17: 40022409-40022409 on Build GRCh38
rs374553629 Chr.17: 40022449-40022449 on Build GRCh38
rs189025204 Chr.17: 40022296-40022296 on Build GRCh38
rs551190459 Chr.17: 40022368-40022368 on Build GRCh38
rs562425968 Chr.17: 40022333-40022333 on Build GRCh38
rs137866788 Chr.17: 40022407-40022407 on Build GRCh38
rs913807148 Chr.17: 40022434-40022434 on Build GRCh38
rs963302181 Chr.17: 40022385-40022385 on Build GRCh38
rs745781592 Chr.17: 40022425-40022425 on Build GRCh38
rs181558536 Chr.17: 40022448-40022448 on Build GRCh38
rs752285482 Chr.17: 40022386-40022386 on Build GRCh38
rs1045277401 Chr.17: 40022441-40022441 on Build GRCh38
rs557148038 Chr.17: 40022433-40022433 on Build GRCh38

View Full Product Details