Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190901856-190902109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS
Entrez Gene ID
2744
Gene Name
glutaminase
Gene Aliases
AAD20, GAC, GAM, GLS1, KGA
UniGene
Hs.116448
Gene Chromosome Location
Chr.2: 190880821-190965552 on Build GRCh38
Transcripts
NM_001256310.1 NM_014905.4 XM_005246467.3 XM_006712435.1 XM_017003829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2712658 c.815G>T p.R272M Chr.2: 190902026-190902026 on Build GRCh38
7135358 c.768C>A p.F256L Chr.2: 190901979-190901979 on Build GRCh38
3743486 c.748A>C p.I250L Chr.2: 190901959-190901959 on Build GRCh38
7135357 c.768C>A p.F256L Chr.2: 190901979-190901979 on Build GRCh38
4308080 c.815G>T p.R272M Chr.2: 190902026-190902026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371550331 Chr.2: 190902054-190902054 on Build GRCh38
rs766626104 Chr.2: 190901947-190901947 on Build GRCh38
rs767267299 Chr.2: 190902028-190902028 on Build GRCh38
rs759901485 Chr.2: 190902074-190902074 on Build GRCh38
rs199780946 Chr.2: 190901944-190901944 on Build GRCh38
rs533233003 Chr.2: 190901913-190901913 on Build GRCh38
rs745616992 Chr.2: 190901910-190901910 on Build GRCh38
rs769525810 Chr.2: 190902029-190902029 on Build GRCh38
rs368638821 Chr.2: 190901921-190901921 on Build GRCh38
rs769497640 Chr.2: 190901912-190901912 on Build GRCh38
rs376240021 Chr.2: 190901906-190901906 on Build GRCh38
rs567565445 Chr.2: 190902080-190902080 on Build GRCh38
rs368025858 Chr.2: 190901969-190901969 on Build GRCh38
rs775538799 Chr.2: 190902058-190902067 on Build GRCh38
rs754152267 Chr.2: 190902070-190902070 on Build GRCh38
rs753222270 Chr.2: 190902077-190902077 on Build GRCh38
rs756770779 Chr.2: 190902006-190902006 on Build GRCh38
rs762767876 Chr.2: 190901916-190901916 on Build GRCh38
rs779464277 Chr.2: 190901985-190901985 on Build GRCh38
rs189809424 Chr.2: 190902083-190902083 on Build GRCh38
rs765677741 Chr.2: 190902075-190902075 on Build GRCh38
rs551776675 Chr.2: 190901917-190901917 on Build GRCh38
rs751006438 Chr.2: 190901986-190901986 on Build GRCh38
rs760786883 Chr.2: 190902064-190902064 on Build GRCh38
rs201068731 Chr.2: 190902044-190902044 on Build GRCh38
rs768709023 Chr.2: 190902047-190902047 on Build GRCh38
rs140570639 Chr.2: 190902017-190902017 on Build GRCh38
rs372566344 Chr.2: 190901923-190901923 on Build GRCh38
rs548107649 Chr.2: 190902056-190902056 on Build GRCh38
rs750699818 Chr.2: 190901942-190901942 on Build GRCh38
rs767793251 Chr.2: 190901929-190901929 on Build GRCh38
rs1020273342 Chr.2: 190901919-190901919 on Build GRCh38
rs16833035 Chr.2: 190901971-190901971 on Build GRCh38
rs142376836 Chr.2: 190901967-190901967 on Build GRCh38
rs780773190 Chr.2: 190902013-190902013 on Build GRCh38
rs779687015 Chr.2: 190902033-190902033 on Build GRCh38
rs911664245 Chr.2: 190902082-190902082 on Build GRCh38

View Full Product Details