Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48750794-48751033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT1
Entrez Gene ID
2523
Gene Name
fucosyltransferase 1 (H blood group)
Gene Aliases
H, HH, HSC
UniGene
Hs.69747
Gene Chromosome Location
Chr.19: 48747826-48755390 on Build GRCh38
Transcripts
NM_000148.3 XM_006723127.2 XM_017026552.1 XM_017026553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395721 c.425G>T p.R142L Chr.19: 48750857-48750857 on Build GRCh38
1395115 c.320G>A p.G107D Chr.19: 48750962-48750962 on Build GRCh38
6667852 c.414C>A p.R138R Chr.19: 48750868-48750868 on Build GRCh38
4981510 c.416C>T p.T139M Chr.19: 48750866-48750866 on Build GRCh38
6667855 c.339G>A p.L113L Chr.19: 48750943-48750943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780921182 Chr.19: 48750910-48750910 on Build GRCh38
rs770949163 Chr.19: 48750863-48750863 on Build GRCh38
rs201562036 Chr.19: 48750864-48750864 on Build GRCh38
rs764415935 Chr.19: 48750819-48750819 on Build GRCh38
rs762089317 Chr.19: 48750871-48750871 on Build GRCh38
rs944826239 Chr.19: 48750967-48750967 on Build GRCh38
rs747599396 Chr.19: 48750846-48750846 on Build GRCh38
rs368762099 Chr.19: 48750894-48750894 on Build GRCh38
rs897674938 Chr.19: 48750903-48750903 on Build GRCh38
rs1023378594 Chr.19: 48750878-48750878 on Build GRCh38
rs780144618 Chr.19: 48750972-48750972 on Build GRCh38
rs781017177 Chr.19: 48750842-48750842 on Build GRCh38
rs749165173 Chr.19: 48750860-48750860 on Build GRCh38
rs752107701 Chr.19: 48750980-48750980 on Build GRCh38
rs775322530 Chr.19: 48750948-48750948 on Build GRCh38
rs760184503 Chr.19: 48750984-48750984 on Build GRCh38
rs56342683 Chr.19: 48750954-48750954 on Build GRCh38
rs765667467 Chr.19: 48750828-48750828 on Build GRCh38
rs765444091 Chr.19: 48750916-48750916 on Build GRCh38
rs772921327 Chr.19: 48750858-48750858 on Build GRCh38
rs754550856 Chr.19: 48750841-48750841 on Build GRCh38
rs781278978 Chr.19: 48750875-48750875 on Build GRCh38
rs764251465 Chr.19: 48750865-48750865 on Build GRCh38
rs146374529 Chr.19: 48750909-48750909 on Build GRCh38
rs561905996 Chr.19: 48750895-48750895 on Build GRCh38
rs779684982 Chr.19: 48750926-48750926 on Build GRCh38
rs777392561 Chr.19: 48750917-48750917 on Build GRCh38
rs897829842 Chr.19: 48751013-48751013 on Build GRCh38
rs776843367 Chr.19: 48750870-48750870 on Build GRCh38
rs529462057 Chr.19: 48750911-48750911 on Build GRCh38
rs750566592 Chr.19: 48750892-48750892 on Build GRCh38
rs769736528 Chr.19: 48750943-48750943 on Build GRCh38
rs757349699 Chr.19: 48750822-48750822 on Build GRCh38
rs769428653 Chr.19: 48750857-48750857 on Build GRCh38
rs144199917 Chr.19: 48750945-48750945 on Build GRCh38
rs765289129 Chr.19: 48750880-48750880 on Build GRCh38
rs747442925 Chr.19: 48750840-48750840 on Build GRCh38
rs970490944 Chr.19: 48750887-48750887 on Build GRCh38
rs758453543 Chr.19: 48750937-48750937 on Build GRCh38
rs527278015 Chr.19: 48750999-48750999 on Build GRCh38
rs753874356 Chr.19: 48750820-48750820 on Build GRCh38
rs139404972 Chr.19: 48750928-48750928 on Build GRCh38
rs751826480 Chr.19: 48750908-48750908 on Build GRCh38
rs773128149 Chr.19: 48750957-48750957 on Build GRCh38
rs750925379 Chr.19: 48750829-48750829 on Build GRCh38
rs745740199 Chr.19: 48750884-48750886 on Build GRCh38
rs778931751 Chr.19: 48750934-48750934 on Build GRCh38
rs753159303 Chr.19: 48751005-48751005 on Build GRCh38
rs763120566 Chr.19: 48750965-48750965 on Build GRCh38
rs762002514 Chr.19: 48750949-48750949 on Build GRCh38
rs759004558 Chr.19: 48750830-48750830 on Build GRCh38
rs150074056 Chr.19: 48750933-48750933 on Build GRCh38
rs932138665 Chr.19: 48750942-48750942 on Build GRCh38
rs780261264 Chr.19: 48750835-48750835 on Build GRCh38

View Full Product Details