Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17142648-17142891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPRIP
Entrez Gene ID
23164
Gene Name
myosin phosphatase Rho interacting protein
Gene Aliases
M-RIP, MRIP, RHOIP3, RIP3, p116Rip
UniGene
Hs.462341
Gene Chromosome Location
Chr.17: 17042438-17192648 on Build GRCh38
Transcripts
NM_015134.3 NM_201274.3 XM_005256563.4 XM_005256564.4 XM_011523761.2 XM_011523762.2 XM_011523763.2 XM_011523764.2 XM_011523765.2 XM_011523766.2 XM_011523767.2 XM_017024393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766013 c.980C>A p.P327Q Chr.17: 17142710-17142710 on Build GRCh38
6322137 c.948G>T p.E316D Chr.17: 17142678-17142678 on Build GRCh38
5571227 c.967G>A p.V323M Chr.17: 17142697-17142697 on Build GRCh38
5851929 c.1034G>A p.R345Q Chr.17: 17142764-17142764 on Build GRCh38
6714910 c.987C>T p.S329S Chr.17: 17142717-17142717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750675265 Chr.17: 17142731-17142731 on Build GRCh38
rs146439071 Chr.17: 17142717-17142717 on Build GRCh38
rs954060809 Chr.17: 17142853-17142853 on Build GRCh38
rs370113289 Chr.17: 17142811-17142811 on Build GRCh38
rs765723394 Chr.17: 17142791-17142791 on Build GRCh38
rs748993550 Chr.17: 17142750-17142750 on Build GRCh38
rs763210949 Chr.17: 17142805-17142805 on Build GRCh38
rs745439092 Chr.17: 17142757-17142757 on Build GRCh38
rs777287779 Chr.17: 17142764-17142764 on Build GRCh38
rs144018217 Chr.17: 17142713-17142713 on Build GRCh38
rs965402783 Chr.17: 17142756-17142756 on Build GRCh38
rs917281191 Chr.17: 17142854-17142854 on Build GRCh38
rs975212127 Chr.17: 17142797-17142797 on Build GRCh38
rs777364536 Chr.17: 17142749-17142749 on Build GRCh38
rs143411460 Chr.17: 17142763-17142763 on Build GRCh38
rs767554583 Chr.17: 17142794-17142795 on Build GRCh38
rs866834193 Chr.17: 17142807-17142807 on Build GRCh38
rs77888743 Chr.17: 17142831-17142831 on Build GRCh38
rs201947495 Chr.17: 17142694-17142694 on Build GRCh38
rs367819704 Chr.17: 17142810-17142810 on Build GRCh38
rs372125214 Chr.17: 17142729-17142729 on Build GRCh38
rs11870441 Chr.17: 17142841-17142841 on Build GRCh38
rs924491641 Chr.17: 17142679-17142679 on Build GRCh38
rs754252940 Chr.17: 17142720-17142720 on Build GRCh38
rs931530654 Chr.17: 17142760-17142760 on Build GRCh38
rs758365889 Chr.17: 17142733-17142733 on Build GRCh38
rs762198610 Chr.17: 17142771-17142771 on Build GRCh38
rs766548826 Chr.17: 17142809-17142809 on Build GRCh38
rs978555370 Chr.17: 17142818-17142818 on Build GRCh38
rs11870488 Chr.17: 17142832-17142832 on Build GRCh38
rs747881775 Chr.17: 17142748-17142748 on Build GRCh38
rs773255014 Chr.17: 17142804-17142804 on Build GRCh38
rs535239889 Chr.17: 17142858-17142858 on Build GRCh38
rs200673181 Chr.17: 17142695-17142695 on Build GRCh38
rs775816038 Chr.17: 17142716-17142716 on Build GRCh38
rs374858004 Chr.17: 17142738-17142738 on Build GRCh38
rs200010089 Chr.17: 17142696-17142696 on Build GRCh38
rs770542092 Chr.17: 17142751-17142751 on Build GRCh38
rs934653040 Chr.17: 17142692-17142692 on Build GRCh38
rs3744137 Chr.17: 17142710-17142710 on Build GRCh38
rs753060959 Chr.17: 17142803-17142813 on Build GRCh38
rs764740780 Chr.17: 17142718-17142718 on Build GRCh38
rs781172214 Chr.17: 17142739-17142739 on Build GRCh38
rs762627702 Chr.17: 17142785-17142786 on Build GRCh38
rs1044857589 Chr.17: 17142697-17142697 on Build GRCh38
rs773885374 Chr.17: 17142755-17142755 on Build GRCh38
rs762323243 Chr.17: 17142725-17142725 on Build GRCh38
rs766502184 Chr.17: 17142734-17142734 on Build GRCh38

View Full Product Details