Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28714605-28714856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3804886 c.1688A>T p.D563V Chr.1: 28714769-28714769 on Build GRCh38
6063073 c.1553C>T p.P518L Chr.1: 28714634-28714634 on Build GRCh38
907998 c.1620G>T p.E540D Chr.1: 28714701-28714701 on Build GRCh38
3487814 c.1687G>A p.V563I Chr.1: 28714738-28714738 on Build GRCh38
4405307 c.1686A>G p.Q562Q Chr.1: 28714737-28714737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771711337 Chr.1: 28714810-28714810 on Build GRCh38
rs780473449 Chr.1: 28714739-28714739 on Build GRCh38
rs779279054 Chr.1: 28714718-28714718 on Build GRCh38
rs143969790 Chr.1: 28714715-28714715 on Build GRCh38
rs868042215 Chr.1: 28714698-28714698 on Build GRCh38
rs775152143 Chr.1: 28714821-28714821 on Build GRCh38
rs781360608 Chr.1: 28714775-28714775 on Build GRCh38
rs758660176 Chr.1: 28714731-28714731 on Build GRCh38
rs750870709 Chr.1: 28714721-28714721 on Build GRCh38
rs760197802 Chr.1: 28714693-28714693 on Build GRCh38
rs368736183 Chr.1: 28714688-28714688 on Build GRCh38
rs760251096 Chr.1: 28714822-28714822 on Build GRCh38
rs202243161 Chr.1: 28714745-28714745 on Build GRCh38
rs748224487 Chr.1: 28714779-28714779 on Build GRCh38
rs773306298 Chr.1: 28714635-28714635 on Build GRCh38
rs755141234 Chr.1: 28714768-28714768 on Build GRCh38
rs774259966 Chr.1: 28714675-28714675 on Build GRCh38
rs148243859 Chr.1: 28714634-28714634 on Build GRCh38
rs16837669 Chr.1: 28714791-28714791 on Build GRCh38
rs745721067 Chr.1: 28714799-28714799 on Build GRCh38
rs570437550 Chr.1: 28714820-28714820 on Build GRCh38
rs767235764 Chr.1: 28714685-28714685 on Build GRCh38
rs150792639 Chr.1: 28714651-28714651 on Build GRCh38
rs758917680 Chr.1: 28714823-28714824 on Build GRCh38
rs978400215 Chr.1: 28714728-28714728 on Build GRCh38
rs747180402 Chr.1: 28714762-28714762 on Build GRCh38
rs770644209 Chr.1: 28714661-28714661 on Build GRCh38
rs763552626 Chr.1: 28714700-28714700 on Build GRCh38
rs774124236 Chr.1: 28714795-28714795 on Build GRCh38
rs754349679 Chr.1: 28714706-28714706 on Build GRCh38
rs746275735 Chr.1: 28714799-28714799 on Build GRCh38
rs759246525 Chr.1: 28714684-28714684 on Build GRCh38

View Full Product Details