Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141100772-141101031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB3
Entrez Gene ID
56132
Gene Name
protocadherin beta 3
Gene Aliases
PCDH-BETA3
UniGene
Hs.508987
Gene Chromosome Location
Chr.5: 141100242-141103827 on Build GRCh38
Transcripts
NM_018937.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
348068 c.158G>T p.G53V Chr.5: 141100807-141100807 on Build GRCh38
6169613 c.130T>C p.L44L Chr.5: 141100779-141100779 on Build GRCh38
1567416 c.349G>A p.V117I Chr.5: 141100998-141100998 on Build GRCh38
3918987 c.180C>T p.A60A Chr.5: 141100829-141100829 on Build GRCh38
1733265 c.164G>T p.R55M Chr.5: 141100813-141100813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551879104 Chr.5: 141100925-141100925 on Build GRCh38
rs149035214 Chr.5: 141100869-141100869 on Build GRCh38
rs374613377 Chr.5: 141100998-141100998 on Build GRCh38
rs782591146 Chr.5: 141100937-141100937 on Build GRCh38
rs782520817 Chr.5: 141100828-141100828 on Build GRCh38
rs782362469 Chr.5: 141100943-141100943 on Build GRCh38
rs782642915 Chr.5: 141100910-141100910 on Build GRCh38
rs140168051 Chr.5: 141100986-141100986 on Build GRCh38
rs782593924 Chr.5: 141100955-141100955 on Build GRCh38
rs782489961 Chr.5: 141100854-141100854 on Build GRCh38
rs781839207 Chr.5: 141100825-141100825 on Build GRCh38
rs782178368 Chr.5: 141100940-141100940 on Build GRCh38
rs781921035 Chr.5: 141100885-141100885 on Build GRCh38
rs782066080 Chr.5: 141100977-141100977 on Build GRCh38
rs782231859 Chr.5: 141100926-141100927 on Build GRCh38
rs151181060 Chr.5: 141100901-141100901 on Build GRCh38
rs943294647 Chr.5: 141100997-141100997 on Build GRCh38
rs782170725 Chr.5: 141100853-141100853 on Build GRCh38
rs201559467 Chr.5: 141100858-141100858 on Build GRCh38
rs782744807 Chr.5: 141100888-141100888 on Build GRCh38
rs781900680 Chr.5: 141100848-141100848 on Build GRCh38
rs782097059 Chr.5: 141100886-141100886 on Build GRCh38
rs782381762 Chr.5: 141100867-141100867 on Build GRCh38
rs782505633 Chr.5: 141100909-141100909 on Build GRCh38
rs782793044 Chr.5: 141100890-141100890 on Build GRCh38
rs61739886 Chr.5: 141100891-141100891 on Build GRCh38
rs782017556 Chr.5: 141100974-141100974 on Build GRCh38
rs200938769 Chr.5: 141100971-141100971 on Build GRCh38
rs782760494 Chr.5: 141100839-141100839 on Build GRCh38
rs782020503 Chr.5: 141100795-141100795 on Build GRCh38
rs782243193 Chr.5: 141100960-141100960 on Build GRCh38
rs782166218 Chr.5: 141100889-141100889 on Build GRCh38
rs782069308 Chr.5: 141100802-141100802 on Build GRCh38
rs782439804 Chr.5: 141100914-141100914 on Build GRCh38
rs782445764 Chr.5: 141100895-141100895 on Build GRCh38
rs935819273 Chr.5: 141100959-141100959 on Build GRCh38
rs782278301 Chr.5: 141100874-141100874 on Build GRCh38
rs782270748 Chr.5: 141100863-141100863 on Build GRCh38
rs782693986 Chr.5: 141100809-141100809 on Build GRCh38
rs782589888 Chr.5: 141100851-141100851 on Build GRCh38
rs781812958 Chr.5: 141100908-141100908 on Build GRCh38
rs782540843 Chr.5: 141100935-141100935 on Build GRCh38
rs17844387 Chr.5: 141100994-141100994 on Build GRCh38
rs146870005 Chr.5: 141100849-141100849 on Build GRCh38

View Full Product Details