Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70297266-70297511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM136A
Entrez Gene ID
84908
Gene Name
family with sequence similarity 136 member A
Gene Aliases
-
UniGene
Hs.516077
Gene Chromosome Location
Chr.2: 70295976-70302088 on Build GRCh38
Transcripts
NM_032822.2 XM_005264620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2152544 c.375G>A p.M125I Chr.2: 70297331-70297331 on Build GRCh38
4164568 c.339G>A p.K113K Chr.2: 70297367-70297367 on Build GRCh38
239773 c.241C>T p.R81W Chr.2: 70297465-70297465 on Build GRCh38
443146 c.399C>T p.L133L Chr.2: 70297307-70297307 on Build GRCh38
6158632 c.259A>T p.N87Y Chr.2: 70297447-70297447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199941425 Chr.2: 70297381-70297381 on Build GRCh38
rs79236443 Chr.2: 70297301-70297301 on Build GRCh38
rs761817680 Chr.2: 70297380-70297383 on Build GRCh38
rs761644349 Chr.2: 70297354-70297354 on Build GRCh38
rs373988641 Chr.2: 70297472-70297472 on Build GRCh38
rs770654285 Chr.2: 70297365-70297365 on Build GRCh38
rs201866753 Chr.2: 70297383-70297383 on Build GRCh38
rs78213014 Chr.2: 70297295-70297295 on Build GRCh38
rs189658423 Chr.2: 70297484-70297484 on Build GRCh38
rs770883166 Chr.2: 70297345-70297345 on Build GRCh38
rs148775834 Chr.2: 70297455-70297455 on Build GRCh38
rs754622369 Chr.2: 70297386-70297386 on Build GRCh38
rs200723862 Chr.2: 70297307-70297307 on Build GRCh38
rs779825246 Chr.2: 70297309-70297309 on Build GRCh38
rs753927903 Chr.2: 70297376-70297377 on Build GRCh38
rs753344193 Chr.2: 70297373-70297373 on Build GRCh38
rs776604234 Chr.2: 70297429-70297429 on Build GRCh38
rs200969130 Chr.2: 70297325-70297326 on Build GRCh38
rs80277652 Chr.2: 70297320-70297320 on Build GRCh38
rs201608469 Chr.2: 70297379-70297379 on Build GRCh38
rs148038860 Chr.2: 70297419-70297419 on Build GRCh38
rs77747732 Chr.2: 70297305-70297305 on Build GRCh38
rs747156050 Chr.2: 70297333-70297333 on Build GRCh38
rs766277089 Chr.2: 70297463-70297463 on Build GRCh38
rs200691537 Chr.2: 70297382-70297382 on Build GRCh38
rs748566832 Chr.2: 70297389-70297389 on Build GRCh38
rs565543670 Chr.2: 70297462-70297462 on Build GRCh38
rs761769037 Chr.2: 70297446-70297446 on Build GRCh38
rs765787726 Chr.2: 70297366-70297366 on Build GRCh38
rs75143374 Chr.2: 70297346-70297346 on Build GRCh38
rs79696069 Chr.2: 70297474-70297474 on Build GRCh38
rs757709605 Chr.2: 70297482-70297482 on Build GRCh38
rs781209587 Chr.2: 70297302-70297302 on Build GRCh38
rs754493203 Chr.2: 70297465-70297465 on Build GRCh38
rs555104422 Chr.2: 70297456-70297456 on Build GRCh38
rs138779159 Chr.2: 70297413-70297413 on Build GRCh38
rs765459159 Chr.2: 70297351-70297351 on Build GRCh38
rs758754729 Chr.2: 70297402-70297402 on Build GRCh38
rs774293245 Chr.2: 70297352-70297352 on Build GRCh38
rs368899781 Chr.2: 70297464-70297464 on Build GRCh38
rs78704688 Chr.2: 70297312-70297312 on Build GRCh38
rs773334625 Chr.2: 70297452-70297452 on Build GRCh38
rs533206742 Chr.2: 70297362-70297362 on Build GRCh38
rs543076072 Chr.2: 70297473-70297473 on Build GRCh38
rs760878746 Chr.2: 70297305-70297306 on Build GRCh38
rs371369005 Chr.2: 70297328-70297328 on Build GRCh38
rs779170909 Chr.2: 70297319-70297319 on Build GRCh38
rs767621848 Chr.2: 70297359-70297359 on Build GRCh38
rs199565792 Chr.2: 70297468-70297468 on Build GRCh38
rs764261137 Chr.2: 70297338-70297338 on Build GRCh38
rs957724915 Chr.2: 70297401-70297401 on Build GRCh38
rs772576584 Chr.2: 70297326-70297326 on Build GRCh38
rs75086964 Chr.2: 70297444-70297444 on Build GRCh38
rs2618235 Chr.2: 70297445-70297445 on Build GRCh38
rs150543829 Chr.2: 70297421-70297421 on Build GRCh38
rs767392518 Chr.2: 70297448-70297448 on Build GRCh38
rs759698708 Chr.2: 70297348-70297348 on Build GRCh38

View Full Product Details