Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102352381-102352638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBF1
Entrez Gene ID
8729
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
Gene Aliases
ARF1GEF
UniGene
Hs.290243
Gene Chromosome Location
Chr.10: 102245498-102382899 on Build GRCh38
Transcripts
NM_001199378.1 NM_001199379.1 NM_004193.2 XM_005270261.1 XM_006718047.1 XM_006718048.1 XM_006718049.1 XM_006718050.1 XM_011540312.1 XM_011540313.1 XM_017016861.1 XM_017016862.1 XM_017016863.1 XM_017016864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682426 c.550A>G p.T184A Chr.10: 102352484-102352484 on Build GRCh38
6453695 c.546G>A p.E182E Chr.10: 102352480-102352480 on Build GRCh38
2059006 c.556G>A p.V186I Chr.10: 102352490-102352490 on Build GRCh38
4500018 c.555C>T p.L185L Chr.10: 102352489-102352489 on Build GRCh38
6128159 c.562A>G p.M188V Chr.10: 102352496-102352496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759772052 Chr.10: 102352533-102352533 on Build GRCh38
rs376175528 Chr.10: 102352490-102352490 on Build GRCh38
rs554640715 Chr.10: 102352604-102352604 on Build GRCh38
rs116123526 Chr.10: 102352451-102352451 on Build GRCh38
rs377584991 Chr.10: 102352583-102352583 on Build GRCh38
rs767823624 Chr.10: 102352566-102352566 on Build GRCh38
rs200460216 Chr.10: 102352568-102352568 on Build GRCh38
rs754650524 Chr.10: 102352475-102352475 on Build GRCh38
rs368601684 Chr.10: 102352516-102352516 on Build GRCh38
rs371872645 Chr.10: 102352542-102352542 on Build GRCh38
rs536655583 Chr.10: 102352570-102352570 on Build GRCh38
rs746983792 Chr.10: 102352508-102352508 on Build GRCh38
rs981150481 Chr.10: 102352543-102352543 on Build GRCh38
rs765668355 Chr.10: 102352448-102352448 on Build GRCh38
rs996740776 Chr.10: 102352507-102352507 on Build GRCh38
rs890957821 Chr.10: 102352581-102352581 on Build GRCh38
rs752730462 Chr.10: 102352420-102352420 on Build GRCh38
rs573305735 Chr.10: 102352616-102352616 on Build GRCh38
rs776588776 Chr.10: 102352527-102352527 on Build GRCh38
rs777558712 Chr.10: 102352501-102352501 on Build GRCh38
rs770808599 Chr.10: 102352526-102352526 on Build GRCh38
rs764287409 Chr.10: 102352547-102352547 on Build GRCh38
rs753451532 Chr.10: 102352474-102352474 on Build GRCh38
rs147305841 Chr.10: 102352606-102352607 on Build GRCh38
rs762340741 Chr.10: 102352556-102352556 on Build GRCh38
rs939559819 Chr.10: 102352400-102352400 on Build GRCh38
rs756568537 Chr.10: 102352562-102352563 on Build GRCh38
rs778430669 Chr.10: 102352487-102352487 on Build GRCh38
rs370884408 Chr.10: 102352452-102352452 on Build GRCh38
rs796350873 Chr.10: 102352562-102352563 on Build GRCh38
rs1038347944 Chr.10: 102352489-102352489 on Build GRCh38
rs576950758 Chr.10: 102352495-102352495 on Build GRCh38
rs893013763 Chr.10: 102352442-102352443 on Build GRCh38
rs775844296 Chr.10: 102352546-102352546 on Build GRCh38
rs957086156 Chr.10: 102352617-102352617 on Build GRCh38
rs545793602 Chr.10: 102352601-102352601 on Build GRCh38
rs1035787188 Chr.10: 102352589-102352589 on Build GRCh38
rs763447542 Chr.10: 102352426-102352426 on Build GRCh38

View Full Product Details