Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43012942-43013215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI35
Entrez Gene ID
3430
Gene Name
interferon induced protein 35
Gene Aliases
IFP35
UniGene
Hs.632258
Gene Chromosome Location
Chr.17: 43006725-43014459 on Build GRCh38
Transcripts
NM_005533.4 XM_005257302.4 XM_017024584.1 XM_017024585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
269347 c.233C>T p.A78V Chr.17: 43013159-43013159 on Build GRCh38
5621028 c.266A>G p.K89R Chr.17: 43013192-43013192 on Build GRCh38
5470680 c.212G>A p.R71Q Chr.17: 43013138-43013138 on Build GRCh38
5157653 c.144delC p.L50fs*17 Chr.17: 43013070-43013070 on Build GRCh38
4989476 c.158G>A p.R53Q Chr.17: 43013084-43013084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770193534 Chr.17: 43013036-43013036 on Build GRCh38
rs932400113 Chr.17: 43012994-43012994 on Build GRCh38
rs879382666 Chr.17: 43012973-43012973 on Build GRCh38
rs765554981 Chr.17: 43012996-43012996 on Build GRCh38
rs780267427 Chr.17: 43013084-43013084 on Build GRCh38
rs148146663 Chr.17: 43012985-43012985 on Build GRCh38
rs771340618 Chr.17: 43013030-43013030 on Build GRCh38
rs756657943 Chr.17: 43013043-43013043 on Build GRCh38
rs187292010 Chr.17: 43013024-43013024 on Build GRCh38
rs36111429 Chr.17: 43013190-43013191 on Build GRCh38
rs772748037 Chr.17: 43013106-43013106 on Build GRCh38
rs200866570 Chr.17: 43013104-43013104 on Build GRCh38
rs758778714 Chr.17: 43013004-43013004 on Build GRCh38
rs778371650 Chr.17: 43013010-43013010 on Build GRCh38
rs751523704 Chr.17: 43013026-43013026 on Build GRCh38
rs779949695 Chr.17: 43013042-43013042 on Build GRCh38
rs368042561 Chr.17: 43013123-43013123 on Build GRCh38
rs781198006 Chr.17: 43013027-43013027 on Build GRCh38
rs768485533 Chr.17: 43013192-43013192 on Build GRCh38
rs1035698617 Chr.17: 43013132-43013133 on Build GRCh38
rs752091266 Chr.17: 43013147-43013147 on Build GRCh38
rs760298021 Chr.17: 43013118-43013118 on Build GRCh38
rs768412142 Chr.17: 43013083-43013083 on Build GRCh38
rs145061445 Chr.17: 43013142-43013142 on Build GRCh38
rs757746781 Chr.17: 43013148-43013148 on Build GRCh38
rs1036850660 Chr.17: 43013111-43013111 on Build GRCh38
rs771605420 Chr.17: 43013105-43013105 on Build GRCh38
rs771604701 Chr.17: 43013145-43013145 on Build GRCh38
rs774360258 Chr.17: 43013102-43013102 on Build GRCh38
rs749034220 Chr.17: 43013185-43013185 on Build GRCh38
rs976426641 Chr.17: 43012980-43012980 on Build GRCh38
rs763377602 Chr.17: 43013138-43013138 on Build GRCh38
rs777520662 Chr.17: 43013160-43013160 on Build GRCh38
rs780428452 Chr.17: 43013174-43013174 on Build GRCh38
rs373561006 Chr.17: 43013079-43013079 on Build GRCh38
rs554750326 Chr.17: 43012992-43012992 on Build GRCh38
rs944132150 Chr.17: 43012974-43012974 on Build GRCh38
rs139469566 Chr.17: 43013137-43013137 on Build GRCh38
rs750862132 Chr.17: 43012986-43012986 on Build GRCh38
rs370391721 Chr.17: 43013066-43013066 on Build GRCh38
rs141921308 Chr.17: 43013159-43013159 on Build GRCh38
rs753109633 Chr.17: 43012997-43012997 on Build GRCh38
rs958627925 Chr.17: 43013162-43013162 on Build GRCh38

View Full Product Details