Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32846557-32846830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPRE1
Entrez Gene ID
22919
Gene Name
microtubule associated protein RP/EB family member 1
Gene Aliases
EB1
UniGene
Hs.472437
Gene Chromosome Location
Chr.20: 32819780-32850405 on Build GRCh38
Transcripts
NM_012325.2 XM_011528696.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6708140 c.603C>T p.N201N Chr.20: 32846623-32846623 on Build GRCh38
3840748 c.633G>A p.E211E Chr.20: 32846653-32846653 on Build GRCh38
6708141 c.655G>A p.G219R Chr.20: 32846675-32846675 on Build GRCh38
4700648 c.654C>T p.F218F Chr.20: 32846674-32846674 on Build GRCh38
6708139 c.712G>A p.V238I Chr.20: 32846732-32846732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776809915 Chr.20: 32846636-32846636 on Build GRCh38
rs370211701 Chr.20: 32846793-32846793 on Build GRCh38
rs765746857 Chr.20: 32846717-32846717 on Build GRCh38
rs375758035 Chr.20: 32846784-32846784 on Build GRCh38
rs751616832 Chr.20: 32846684-32846684 on Build GRCh38
rs767472263 Chr.20: 32846713-32846713 on Build GRCh38
rs377582239 Chr.20: 32846590-32846590 on Build GRCh38
rs746012543 Chr.20: 32846782-32846782 on Build GRCh38
rs749385659 Chr.20: 32846785-32846785 on Build GRCh38
rs769723396 Chr.20: 32846638-32846638 on Build GRCh38
rs142905797 Chr.20: 32846734-32846734 on Build GRCh38
rs878861719 Chr.20: 32846662-32846662 on Build GRCh38
rs41289870 Chr.20: 32846622-32846622 on Build GRCh38
rs928584069 Chr.20: 32846699-32846699 on Build GRCh38
rs1049328712 Chr.20: 32846606-32846606 on Build GRCh38
rs770068452 Chr.20: 32846783-32846783 on Build GRCh38
rs757567678 Chr.20: 32846754-32846754 on Build GRCh38
rs896136714 Chr.20: 32846668-32846668 on Build GRCh38
rs142439935 Chr.20: 32846623-32846623 on Build GRCh38
rs755792170 Chr.20: 32846716-32846716 on Build GRCh38
rs374283932 Chr.20: 32846725-32846725 on Build GRCh38
rs537073299 Chr.20: 32846789-32846789 on Build GRCh38
rs202221243 Chr.20: 32846641-32846641 on Build GRCh38
rs940573329 Chr.20: 32846614-32846614 on Build GRCh38
rs1056327244 Chr.20: 32846644-32846644 on Build GRCh38
rs755560140 Chr.20: 32846596-32846596 on Build GRCh38
rs139716054 Chr.20: 32846686-32846686 on Build GRCh38
rs748534756 Chr.20: 32846605-32846605 on Build GRCh38
rs951830240 Chr.20: 32846603-32846603 on Build GRCh38
rs75036289 Chr.20: 32846800-32846800 on Build GRCh38
rs371221771 Chr.20: 32846714-32846714 on Build GRCh38
rs149878687 Chr.20: 32846801-32846801 on Build GRCh38
rs778221336 Chr.20: 32846732-32846732 on Build GRCh38
rs759774837 Chr.20: 32846637-32846637 on Build GRCh38
rs771324465 Chr.20: 32846624-32846624 on Build GRCh38
rs981306242 Chr.20: 32846650-32846650 on Build GRCh38
rs762741175 Chr.20: 32846674-32846674 on Build GRCh38
rs200062626 Chr.20: 32846639-32846639 on Build GRCh38
rs145958476 Chr.20: 32846683-32846683 on Build GRCh38
rs112403438 Chr.20: 32846612-32846612 on Build GRCh38
rs369653826 Chr.20: 32846598-32846598 on Build GRCh38
rs781569841 Chr.20: 32846768-32846768 on Build GRCh38
rs775645885 Chr.20: 32846645-32846645 on Build GRCh38
rs754509909 Chr.20: 32846726-32846726 on Build GRCh38
rs750428174 Chr.20: 32846786-32846788 on Build GRCh38

View Full Product Details