Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89248834-89249069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN3
Entrez Gene ID
285513
Gene Name
GPRIN family member 3
Gene Aliases
GRIN3
UniGene
Hs.100912
Gene Chromosome Location
Chr.4: 89237666-89308010 on Build GRCh38
Transcripts
NM_198281.2 XM_005262936.3 XM_005262937.4 XM_017008043.1 XM_017008044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1310375 c.1098G>A p.G366G Chr.4: 89249013-89249013 on Build GRCh38
1285258 c.1179C>G p.H393Q Chr.4: 89248932-89248932 on Build GRCh38
3702651 c.1253T>G p.L418R Chr.4: 89248858-89248858 on Build GRCh38
5975270 c.1272T>C p.N424N Chr.4: 89248839-89248839 on Build GRCh38
1131000 c.1144T>C p.S382P Chr.4: 89248967-89248967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762344002 Chr.4: 89248988-89248988 on Build GRCh38
rs752685664 Chr.4: 89249024-89249024 on Build GRCh38
rs755271835 Chr.4: 89249013-89249013 on Build GRCh38
rs375925089 Chr.4: 89248983-89248983 on Build GRCh38
rs774887362 Chr.4: 89248985-89248985 on Build GRCh38
rs551630860 Chr.4: 89248876-89248876 on Build GRCh38
rs772901919 Chr.4: 89248885-89248885 on Build GRCh38
rs758550764 Chr.4: 89249033-89249033 on Build GRCh38
rs778871750 Chr.4: 89248960-89248960 on Build GRCh38
rs748038123 Chr.4: 89248879-89248879 on Build GRCh38
rs201902685 Chr.4: 89248993-89248993 on Build GRCh38
rs760333108 Chr.4: 89248886-89248886 on Build GRCh38
rs141576652 Chr.4: 89248947-89248947 on Build GRCh38
rs532134284 Chr.4: 89248867-89248867 on Build GRCh38
rs113722378 Chr.4: 89248941-89248941 on Build GRCh38
rs35246727 Chr.4: 89248895-89248895 on Build GRCh38
rs35241214 Chr.4: 89248879-89248880 on Build GRCh38
rs755040645 Chr.4: 89248956-89248956 on Build GRCh38
rs750603293 Chr.4: 89248863-89248863 on Build GRCh38
rs1024549870 Chr.4: 89248928-89248928 on Build GRCh38
rs1039076735 Chr.4: 89248865-89248865 on Build GRCh38
rs144396875 Chr.4: 89249019-89249019 on Build GRCh38
rs781417158 Chr.4: 89248862-89248862 on Build GRCh38
rs769292858 Chr.4: 89248984-89248984 on Build GRCh38
rs766855005 Chr.4: 89249008-89249008 on Build GRCh38
rs11733183 Chr.4: 89248943-89248943 on Build GRCh38
rs370470960 Chr.4: 89248870-89248870 on Build GRCh38
rs377675746 Chr.4: 89248878-89248878 on Build GRCh38
rs76496030 Chr.4: 89248946-89248946 on Build GRCh38
rs6811370 Chr.4: 89248978-89248978 on Build GRCh38
rs767876043 Chr.4: 89248935-89248935 on Build GRCh38
rs759828351 Chr.4: 89248880-89248880 on Build GRCh38
rs28622301 Chr.4: 89248967-89248967 on Build GRCh38
rs560428765 Chr.4: 89248894-89248894 on Build GRCh38
rs777554551 Chr.4: 89249035-89249035 on Build GRCh38
rs761115241 Chr.4: 89248997-89248997 on Build GRCh38
rs777802382 Chr.4: 89248969-89248969 on Build GRCh38
rs756983246 Chr.4: 89249046-89249046 on Build GRCh38
rs369882222 Chr.4: 89248992-89248992 on Build GRCh38
rs886607652 Chr.4: 89248915-89248915 on Build GRCh38
rs978358637 Chr.4: 89249000-89249000 on Build GRCh38
rs539048508 Chr.4: 89249036-89249036 on Build GRCh38
rs373856944 Chr.4: 89248903-89248903 on Build GRCh38
rs754054389 Chr.4: 89249010-89249010 on Build GRCh38
rs754179495 Chr.4: 89248948-89248948 on Build GRCh38
rs777576353 Chr.4: 89248937-89248937 on Build GRCh38
rs752040251 Chr.4: 89248919-89248919 on Build GRCh38
rs769101447 Chr.4: 89248879-89248909 on Build GRCh38
rs114960901 Chr.4: 89248864-89248864 on Build GRCh38
rs762242977 Chr.4: 89248931-89248931 on Build GRCh38
rs768913675 Chr.4: 89248871-89248871 on Build GRCh38

View Full Product Details