Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57053872-57054097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1586
Entrez Gene ID
57691
Gene Name
KIAA1586
Gene Aliases
-
UniGene
Hs.709521
Gene Chromosome Location
Chr.6: 57046532-57055739 on Build GRCh38
Transcripts
NM_001286274.1 NM_001286275.1 NM_001286276.1 NM_020931.3 XM_011514767.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3159676 c.1442G>T p.G481V Chr.6: 57053941-57053941 on Build GRCh38
3875014 c.1507T>C p.Y503H Chr.6: 57054006-57054006 on Build GRCh38
6174823 c.1522C>A p.H508N Chr.6: 57054021-57054021 on Build GRCh38
5694842 c.1395A>C p.V465V Chr.6: 57053894-57053894 on Build GRCh38
6238752 c.1451G>T p.W484L Chr.6: 57053950-57053950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147271286 Chr.6: 57054057-57054057 on Build GRCh38
rs777041437 Chr.6: 57054040-57054040 on Build GRCh38
rs767756139 Chr.6: 57053954-57053954 on Build GRCh38
rs375554795 Chr.6: 57053953-57053953 on Build GRCh38
rs760946035 Chr.6: 57054063-57054063 on Build GRCh38
rs781009106 Chr.6: 57054004-57054004 on Build GRCh38
rs368816563 Chr.6: 57053981-57053981 on Build GRCh38
rs751754390 Chr.6: 57054027-57054028 on Build GRCh38
rs752866550 Chr.6: 57053957-57053957 on Build GRCh38
rs370896018 Chr.6: 57053931-57053931 on Build GRCh38
rs201387216 Chr.6: 57053989-57053989 on Build GRCh38
rs764785876 Chr.6: 57053913-57053913 on Build GRCh38
rs772833430 Chr.6: 57054012-57054012 on Build GRCh38
rs1050259882 Chr.6: 57053982-57053982 on Build GRCh38
rs779260527 Chr.6: 57053996-57053996 on Build GRCh38
rs762345177 Chr.6: 57054013-57054013 on Build GRCh38
rs781180579 Chr.6: 57054067-57054068 on Build GRCh38
rs751835703 Chr.6: 57054041-57054041 on Build GRCh38
rs763121707 Chr.6: 57053993-57053993 on Build GRCh38
rs866707605 Chr.6: 57054021-57054021 on Build GRCh38
rs757295403 Chr.6: 57054060-57054060 on Build GRCh38
rs761434486 Chr.6: 57053904-57053904 on Build GRCh38
rs751416959 Chr.6: 57053948-57053948 on Build GRCh38
rs772520152 Chr.6: 57054003-57054003 on Build GRCh38
rs143927840 Chr.6: 57054031-57054031 on Build GRCh38
rs993487357 Chr.6: 57054019-57054019 on Build GRCh38
rs766210734 Chr.6: 57053932-57053932 on Build GRCh38
rs925828264 Chr.6: 57053969-57053969 on Build GRCh38
rs375040639 Chr.6: 57054009-57054009 on Build GRCh38
rs777897727 Chr.6: 57053987-57053987 on Build GRCh38
rs777834468 Chr.6: 57054026-57054026 on Build GRCh38
rs371793598 Chr.6: 57054000-57054000 on Build GRCh38
rs747731774 Chr.6: 57054006-57054006 on Build GRCh38

View Full Product Details