Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 124970615-124970840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEG1
Entrez Gene ID
57493
Gene Name
heart development protein with EGF like domains 1
Gene Aliases
HEG, MST112, MSTP112
UniGene
Hs.477420
Gene Chromosome Location
Chr.3: 124965710-125055958 on Build GRCh38
Transcripts
NM_020733.1 XM_005247666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261255 c.4110C>A p.F1370L Chr.3: 124970688-124970688 on Build GRCh38
257101 c.4028G>A p.R1343Q Chr.3: 124970770-124970770 on Build GRCh38
6678960 c.4104G>A p.P1368P Chr.3: 124970694-124970694 on Build GRCh38
4546842 c.4090G>T p.G1364* Chr.3: 124970708-124970708 on Build GRCh38
5863386 c.4044_4045GG>AA p.A1349T Chr.3: 124970753-124970754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747834813 Chr.3: 124970728-124970728 on Build GRCh38
rs757079640 Chr.3: 124970658-124970658 on Build GRCh38
rs374923347 Chr.3: 124970694-124970694 on Build GRCh38
rs751775083 Chr.3: 124970771-124970771 on Build GRCh38
rs765669476 Chr.3: 124970755-124970755 on Build GRCh38
rs936332912 Chr.3: 124970757-124970757 on Build GRCh38
rs770547898 Chr.3: 124970800-124970800 on Build GRCh38
rs371908358 Chr.3: 124970639-124970639 on Build GRCh38
rs764112615 Chr.3: 124970770-124970770 on Build GRCh38
rs990138256 Chr.3: 124970746-124970746 on Build GRCh38
rs775877355 Chr.3: 124970765-124970765 on Build GRCh38
rs748772887 Chr.3: 124970683-124970683 on Build GRCh38
rs1018362600 Chr.3: 124970674-124970674 on Build GRCh38
rs923094201 Chr.3: 124970701-124970701 on Build GRCh38
rs1054759888 Chr.3: 124970760-124970760 on Build GRCh38
rs774013058 Chr.3: 124970709-124970709 on Build GRCh38
rs764322121 Chr.3: 124970635-124970635 on Build GRCh38
rs779858815 Chr.3: 124970671-124970671 on Build GRCh38
rs771259089 Chr.3: 124970729-124970729 on Build GRCh38
rs767415348 Chr.3: 124970783-124970783 on Build GRCh38
rs977638748 Chr.3: 124970672-124970672 on Build GRCh38
rs375218561 Chr.3: 124970784-124970784 on Build GRCh38
rs781173052 Chr.3: 124970661-124970661 on Build GRCh38
rs757602890 Chr.3: 124970782-124970782 on Build GRCh38
rs965491106 Chr.3: 124970687-124970687 on Build GRCh38
rs777043322 Chr.3: 124970737-124970737 on Build GRCh38
rs764062830 Chr.3: 124970664-124970666 on Build GRCh38
rs756186979 Chr.3: 124970668-124970668 on Build GRCh38
rs922426827 Chr.3: 124970786-124970786 on Build GRCh38
rs186255239 Chr.3: 124970754-124970754 on Build GRCh38
rs553906393 Chr.3: 124970801-124970801 on Build GRCh38
rs745787138 Chr.3: 124970664-124970664 on Build GRCh38
rs755985532 Chr.3: 124970799-124970799 on Build GRCh38
rs371494538 Chr.3: 124970766-124970766 on Build GRCh38
rs546708760 Chr.3: 124970695-124970695 on Build GRCh38

View Full Product Details