Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1597859-1598100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-2
Entrez Gene ID
440021
Gene Name
keratin associated protein 5-2
Gene Aliases
KRTAP5-8, KRTAP5.2
UniGene
Hs.633229
Gene Chromosome Location
Chr.11: 1597177-1598294 on Build GRCh38
Transcripts
NM_001004325.1 NR_021489.1

Gene Symbol
KRTAP5-AS1
Entrez Gene ID
338651
Gene Name
KRTAP5-1/KRTAP5-2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.633229
Gene Chromosome Location
Chr.11: 1572741-1599184 on Build GRCh38
Transcripts
NM_001004325.1 NR_021489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5263317 c.288G>A p.G96G Chr.11: 1597963-1597963 on Build GRCh38
3415717 c.170A>G p.Y57C Chr.11: 1598081-1598081 on Build GRCh38
4994951 c.392C>A p.S131Y Chr.11: 1597859-1597859 on Build GRCh38
5021700 c.249C>T p.G83G Chr.11: 1598002-1598002 on Build GRCh38
6369374 c.163A>G p.R55G Chr.11: 1598088-1598088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779850306 Chr.11: 1597972-1597997 on Build GRCh38
rs764593718 Chr.11: 1598062-1598062 on Build GRCh38
rs760032721 Chr.11: 1597961-1597966 on Build GRCh38
rs368694018 Chr.11: 1597919-1597919 on Build GRCh38
rs758664201 Chr.11: 1597964-1597968 on Build GRCh38
rs376517981 Chr.11: 1597884-1597884 on Build GRCh38
rs749460852 Chr.11: 1597991-1597991 on Build GRCh38
rs779889964 Chr.11: 1597909-1597909 on Build GRCh38
rs745945703 Chr.11: 1597970-1597994 on Build GRCh38
rs761766394 Chr.11: 1597922-1597922 on Build GRCh38
rs770510497 Chr.11: 1598048-1598048 on Build GRCh38
rs773961574 Chr.11: 1597951-1597951 on Build GRCh38
rs766975121 Chr.11: 1597954-1597955 on Build GRCh38
rs774163393 Chr.11: 1597986-1598013 on Build GRCh38
rs756080173 Chr.11: 1597988-1597988 on Build GRCh38
rs753477733 Chr.11: 1597946-1597946 on Build GRCh38
rs768562801 Chr.11: 1597948-1597948 on Build GRCh38
rs772668808 Chr.11: 1598035-1598035 on Build GRCh38
rs200646523 Chr.11: 1597993-1597993 on Build GRCh38
rs748081167 Chr.11: 1598007-1598007 on Build GRCh38
rs746117231 Chr.11: 1597902-1597902 on Build GRCh38
rs772751865 Chr.11: 1597928-1597928 on Build GRCh38
rs982228957 Chr.11: 1598033-1598033 on Build GRCh38
rs61869706 Chr.11: 1597972-1597972 on Build GRCh38
rs771727043 Chr.11: 1597998-1598009 on Build GRCh38
rs915735520 Chr.11: 1598068-1598068 on Build GRCh38
rs757580577 Chr.11: 1597968-1597995 on Build GRCh38
rs938514531 Chr.11: 1598056-1598056 on Build GRCh38
rs780266459 Chr.11: 1597989-1597989 on Build GRCh38
rs765147746 Chr.11: 1598002-1598002 on Build GRCh38
rs961671053 Chr.11: 1598030-1598030 on Build GRCh38
rs752145293 Chr.11: 1597966-1597966 on Build GRCh38
rs772977729 Chr.11: 1597998-1598005 on Build GRCh38
rs765957533 Chr.11: 1598032-1598053 on Build GRCh38
rs747734464 Chr.11: 1597990-1597991 on Build GRCh38
rs531448786 Chr.11: 1597897-1597897 on Build GRCh38
rs61869704 Chr.11: 1597951-1597951 on Build GRCh38
rs36134435 Chr.11: 1597954-1597954 on Build GRCh38
rs760241097 Chr.11: 1598045-1598045 on Build GRCh38
rs749288386 Chr.11: 1597973-1597994 on Build GRCh38
rs751779991 Chr.11: 1597968-1597991 on Build GRCh38
rs142577062 Chr.11: 1597933-1597933 on Build GRCh38
rs549555085 Chr.11: 1597912-1597912 on Build GRCh38
rs374648335 Chr.11: 1597901-1597901 on Build GRCh38
rs781352206 Chr.11: 1597969-1597996 on Build GRCh38
rs753317529 Chr.11: 1597962-1597990 on Build GRCh38
rs770043929 Chr.11: 1597906-1597906 on Build GRCh38
rs370177579 Chr.11: 1597888-1597888 on Build GRCh38
rs757753042 Chr.11: 1597967-1597967 on Build GRCh38
rs749942570 Chr.11: 1597961-1597964 on Build GRCh38
rs908577919 Chr.11: 1597935-1597935 on Build GRCh38
rs750511429 Chr.11: 1597987-1597987 on Build GRCh38
rs756273632 Chr.11: 1597970-1597971 on Build GRCh38
rs547259506 Chr.11: 1597973-1597973 on Build GRCh38
rs199811378 Chr.11: 1597963-1597963 on Build GRCh38
rs59506446 Chr.11: 1597992-1598021 on Build GRCh38
rs376100963 Chr.11: 1598055-1598055 on Build GRCh38
rs765652724 Chr.11: 1597961-1597962 on Build GRCh38
rs759225655 Chr.11: 1598058-1598058 on Build GRCh38
rs771519063 Chr.11: 1597925-1597925 on Build GRCh38
rs759044515 Chr.11: 1597895-1597895 on Build GRCh38
rs768392223 Chr.11: 1597979-1597996 on Build GRCh38
rs766128627 Chr.11: 1597936-1597936 on Build GRCh38
rs778335207 Chr.11: 1597968-1597993 on Build GRCh38
rs137933021 Chr.11: 1598013-1598013 on Build GRCh38
rs760014834 Chr.11: 1597943-1597972 on Build GRCh38
rs762640881 Chr.11: 1597925-1597954 on Build GRCh38
rs760101317 Chr.11: 1598009-1598010 on Build GRCh38
rs142046547 Chr.11: 1598026-1598026 on Build GRCh38
rs778644592 Chr.11: 1597998-1597998 on Build GRCh38
rs928001486 Chr.11: 1598043-1598043 on Build GRCh38
rs761477762 Chr.11: 1597954-1597982 on Build GRCh38
rs768468038 Chr.11: 1597995-1597995 on Build GRCh38
rs538549244 Chr.11: 1597942-1597942 on Build GRCh38
rs757307800 Chr.11: 1597900-1597900 on Build GRCh38

View Full Product Details