Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143876041-143876302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAF1
Entrez Gene ID
9747
Gene Name
TRPM8 channel associated factor 1
Gene Aliases
FAM115A
UniGene
Hs.735149
Gene Chromosome Location
Chr.7: 143851368-143902228 on Build GRCh38
Transcripts
NM_001206938.1 NM_001206941.1 NM_014719.2 XM_005250074.3 XM_005250075.2 XM_005250076.3 XM_005250077.2 XM_006716191.2 XM_006716192.3 XM_017012833.1 XM_017012834.1 XM_017012835.1 XM_017012836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1086825 c.344A>G p.E115G Chr.7: 143876265-143876265 on Build GRCh38
5949049 c.548_552delTTACT p.F183fs*1 Chr.7: 143876057-143876061 on Build GRCh38
5997307 c.335C>T p.A112V Chr.7: 143876274-143876274 on Build GRCh38
3878981 c.438C>T p.G146G Chr.7: 143876171-143876171 on Build GRCh38
1086827 c.315C>T p.L105L Chr.7: 143876294-143876294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149240169 Chr.7: 143876093-143876093 on Build GRCh38
rs531046645 Chr.7: 143876254-143876254 on Build GRCh38
rs758300611 Chr.7: 143876068-143876068 on Build GRCh38
rs778944649 Chr.7: 143876139-143876139 on Build GRCh38
rs780254168 Chr.7: 143876087-143876087 on Build GRCh38
rs145611193 Chr.7: 143876185-143876185 on Build GRCh38
rs777359713 Chr.7: 143876114-143876114 on Build GRCh38
rs147747470 Chr.7: 143876209-143876209 on Build GRCh38
rs558859540 Chr.7: 143876135-143876135 on Build GRCh38
rs542994466 Chr.7: 143876269-143876269 on Build GRCh38
rs770632783 Chr.7: 143876129-143876129 on Build GRCh38
rs1014091006 Chr.7: 143876205-143876205 on Build GRCh38
rs774253614 Chr.7: 143876273-143876273 on Build GRCh38
rs766445064 Chr.7: 143876279-143876279 on Build GRCh38
rs762821957 Chr.7: 143876160-143876160 on Build GRCh38
rs745456904 Chr.7: 143876241-143876241 on Build GRCh38
rs200784338 Chr.7: 143876170-143876170 on Build GRCh38
rs541966250 Chr.7: 143876208-143876208 on Build GRCh38
rs370517812 Chr.7: 143876197-143876197 on Build GRCh38
rs373767341 Chr.7: 143876217-143876217 on Build GRCh38
rs150457102 Chr.7: 143876171-143876171 on Build GRCh38
rs775155191 Chr.7: 143876159-143876159 on Build GRCh38
rs779164638 Chr.7: 143876102-143876102 on Build GRCh38
rs144423231 Chr.7: 143876103-143876103 on Build GRCh38
rs554165667 Chr.7: 143876079-143876079 on Build GRCh38
rs1017121326 Chr.7: 143876256-143876256 on Build GRCh38
rs78357950 Chr.7: 143876229-143876229 on Build GRCh38
rs1056343293 Chr.7: 143876239-143876239 on Build GRCh38
rs753871009 Chr.7: 143876283-143876283 on Build GRCh38
rs768475979 Chr.7: 143876261-143876261 on Build GRCh38
rs766415083 Chr.7: 143876174-143876174 on Build GRCh38
rs780596712 Chr.7: 143876240-143876240 on Build GRCh38
rs761765487 Chr.7: 143876275-143876275 on Build GRCh38
rs769342310 Chr.7: 143876247-143876247 on Build GRCh38
rs117822670 Chr.7: 143876105-143876105 on Build GRCh38
rs753014947 Chr.7: 143876201-143876201 on Build GRCh38
rs367932760 Chr.7: 143876225-143876225 on Build GRCh38
rs767520997 Chr.7: 143876075-143876075 on Build GRCh38
rs746683154 Chr.7: 143876210-143876210 on Build GRCh38
rs754039367 Chr.7: 143876184-143876184 on Build GRCh38
rs775267886 Chr.7: 143876249-143876249 on Build GRCh38
rs371535446 Chr.7: 143876162-143876162 on Build GRCh38
rs755343930 Chr.7: 143876097-143876097 on Build GRCh38
rs759134072 Chr.7: 143876133-143876133 on Build GRCh38
rs776324681 Chr.7: 143876132-143876132 on Build GRCh38
rs756247852 Chr.7: 143876086-143876086 on Build GRCh38
rs746503679 Chr.7: 143876128-143876128 on Build GRCh38
rs917822594 Chr.7: 143876270-143876270 on Build GRCh38

View Full Product Details