Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41467997-41468270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF5A
Entrez Gene ID
84844
Gene Name
PHD finger protein 5A
Gene Aliases
INI, Rds3, SAP14b, SF3B7, SF3b14b, bK223H9.2
UniGene
Hs.474980
Gene Chromosome Location
Chr.22: 41459717-41468704 on Build GRCh38
Transcripts
NM_032758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034632 c.69T>C p.C23C Chr.22: 41468131-41468131 on Build GRCh38
292435 c.72A>C p.E24D Chr.22: 41468128-41468128 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778752488 Chr.22: 41468093-41468093 on Build GRCh38
rs746982843 Chr.22: 41468121-41468121 on Build GRCh38
rs773446518 Chr.22: 41468170-41468170 on Build GRCh38
rs998513919 Chr.22: 41468030-41468030 on Build GRCh38
rs768811022 Chr.22: 41468066-41468066 on Build GRCh38
rs757791444 Chr.22: 41468197-41468197 on Build GRCh38
rs774351024 Chr.22: 41468182-41468182 on Build GRCh38
rs373583942 Chr.22: 41468115-41468115 on Build GRCh38
rs141558065 Chr.22: 41468080-41468080 on Build GRCh38
rs532686270 Chr.22: 41468051-41468051 on Build GRCh38
rs888970608 Chr.22: 41468207-41468207 on Build GRCh38
rs768536936 Chr.22: 41468134-41468134 on Build GRCh38
rs968375968 Chr.22: 41468026-41468026 on Build GRCh38
rs745320431 Chr.22: 41468101-41468101 on Build GRCh38
rs753177987 Chr.22: 41468073-41468073 on Build GRCh38
rs776864965 Chr.22: 41468071-41468071 on Build GRCh38
rs570956260 Chr.22: 41468200-41468200 on Build GRCh38
rs505533 Chr.22: 41468212-41468212 on Build GRCh38
rs760246074 Chr.22: 41468200-41468202 on Build GRCh38
rs762903146 Chr.22: 41468171-41468171 on Build GRCh38
rs914757948 Chr.22: 41468154-41468154 on Build GRCh38
rs190531455 Chr.22: 41468108-41468108 on Build GRCh38
rs567184011 Chr.22: 41468163-41468163 on Build GRCh38
rs9611597 Chr.22: 41468186-41468186 on Build GRCh38
rs61736535 Chr.22: 41468048-41468048 on Build GRCh38
rs1042861767 Chr.22: 41468225-41468225 on Build GRCh38
rs200181022 Chr.22: 41468110-41468110 on Build GRCh38
rs548572822 Chr.22: 41468161-41468161 on Build GRCh38
rs534183600 Chr.22: 41468177-41468177 on Build GRCh38
rs555827738 Chr.22: 41468232-41468232 on Build GRCh38
rs970175666 Chr.22: 41468109-41468109 on Build GRCh38
rs904270360 Chr.22: 41468244-41468244 on Build GRCh38
rs1005163343 Chr.22: 41468042-41468042 on Build GRCh38
rs916075574 Chr.22: 41468105-41468105 on Build GRCh38
rs777359697 Chr.22: 41468116-41468116 on Build GRCh38
rs993039626 Chr.22: 41468090-41468090 on Build GRCh38
rs886183671 Chr.22: 41468022-41468022 on Build GRCh38
rs760082051 Chr.22: 41468184-41468184 on Build GRCh38
rs753089765 Chr.22: 41468188-41468188 on Build GRCh38
rs764939173 Chr.22: 41468196-41468196 on Build GRCh38
rs187473076 Chr.22: 41468056-41468056 on Build GRCh38
rs370916619 Chr.22: 41468192-41468192 on Build GRCh38
rs772921041 Chr.22: 41468097-41468099 on Build GRCh38
rs776409922 Chr.22: 41468143-41468143 on Build GRCh38

View Full Product Details